    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PEX6 Antibodies
          Antibodies Inc
          PEX6 Monoclonal Antibody (N233/8), FL594
          View all (11) PEX6 antibodies
          Tech Support
          Cite PEX6 Monoclonal Antibody (N233/8), FL594
          Additional Information:
          {{banner.description}}
          PEX6 Monoclonal Antibody (N233/8), FL594
          Product Details
          75-285-FL594
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N233/8
          Immunogen
          Fusion protein amino acids 2-294 (N-terminus) of human PEX6 (accession number Q8BR86-3) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          FL594 FL594 FL594
          • Unconjugated
          • FL490
          • FL550
          • FL650
          • Request custom conjugation
          Excitation/Emission Max
          594/615 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          PEX6 encodes a member of the AAA (ATPases associated with diverse cellular activities) family of ATPases. This member is a predominantly cytoplasmic protein, which plays a direct role in peroxisomal protein import and is required for PTS1 (peroxisomal targeting signal 1, a C-terminal tripeptide of the sequence ser-lys-leu) receptor activity. Mutations in this gene cause peroxisome biogenesis disorders of complementation group 4 and complementation group 6.
          Bioinformatics
          Protein Aliases: PAF 2; PAF-2; Peroxin 6; Peroxin-6; peroxisomal AAA-type ATPase 1; Peroxisomal ATPase PEX6; Peroxisomal biogenesis factor 6; Peroxisomal-type ATPase 1; Peroxisome assembly factor 2; RP1-125M3.3; unnamed protein product
          View more View less
          Gene Aliases: HMLR2; PAF-2; PAF2; PBD4A; PDB4B; PEX6; PXAAA1
          View more View less
          UniProt ID: (Human) Q13608, (Rat) P54777
          View more View less
          Entrez Gene ID: (Human) 5190, (Rat) 117265
          View more View less
          Function(s)
          nucleotide binding protein binding ATP binding hydrolase activity ATPase activity macromolecular complex binding ubiquitin-dependent protein binding protein transporter activity protein C-terminus binding protein complex binding ATPase activity, coupled
          Process(es)
          protein targeting to peroxisome peroxisome organization metabolic process protein import into peroxisome matrix, translocation protein stabilization protein import into peroxisome matrix protein import into peroxisome matrix, receptor recycling protein unfolding
          France flag icon
          France
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline