    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PRRT1 Antibodies
          Antibodies Inc
          SynDIG4 Monoclonal Antibody (L102/45), FL650
          View all (10) PRRT1 antibodies
          Tech Support
          Cite SynDIG4 Monoclonal Antibody (L102/45), FL650
          Additional Information:
          {{banner.description}}
          SynDIG4 Monoclonal Antibody (L102/45), FL650
          Product Details
          75-409-FL650
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG2
          Class
          Monoclonal
          Type
          Antibody
          Clone
          L102/45
          Immunogen
          Fusion protein amino acids 1-66 (extracellular N-terminus) of rat SynDIG4 (accession number Q6MG82) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          FL650 FL650 FL650
          • Unconjugated
          • FL490
          • FL550
          • FL594
          • Request custom conjugation
          Excitation/Emission Max
          655/676 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          PRRT1 is a protein coding gene.
          Bioinformatics
          Protein Aliases: Dispanin subfamily D member 1; DSPD1; interferon induced transmembrane protein domain containing 7; Proline-rich transmembrane protein 1; prrt1 {ECO:0000250|UniProtKB:Q99946}; Synapse differentiation-induced protein 4; SynDIG4; unnamed protein product
          View more View less
          Gene Aliases: Bat-4; Bat4; C6orf31; D17H6S54E; DSPD1; G5b; IFITMD7; NG5; ORF31; PRRT1; SynDIG4
          View more View less
          UniProt ID: (Human) Q99946, (Rat) Q6MG82, (Mouse) O35449
          View more View less
          Entrez Gene ID: (Human) 80863, (Rat) 406167, (Mouse) 260297
          View more View less
          Function(s)
          protein binding receptor regulator activity identical protein binding molecular_function
          Process(es)
          protein phosphorylation learning or memory protein localization protein localization to cell surface synapse organization long-term synaptic potentiation long term synaptic depression regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity biological_process response to biotic stimulus
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline