    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • ANO5 Antibodies
          Antibodies Inc
          ANO5/TMEM16E Monoclonal Antibody (N421A/85), FL650
          View all (10) ANO5 antibodies
          Tech Support
          Cite ANO5/TMEM16E Monoclonal Antibody (N421A/85), FL650
          Additional Information:
          {{banner.description}}
          ANO5/TMEM16E Monoclonal Antibody (N421A/85), FL650
          Product Details
          75-410-FL650
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N421A/85
          Immunogen
          Fusion protein amino acids 1-299 (cytoplasmic N-terminus) of human ANO5 (accession number Q75V66) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          FL650 FL650 FL650
          • Unconjugated
          • FL490
          • FL550
          • FL594
          • Request custom conjugation
          Excitation/Emission Max
          655/676 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          Ano5 does not exhibit calcium-activated chloride channel (CaCC) activity. The Ano5 gene encodes a member of the anoctamin family of transmembrane proteins. The Ano5 protein is likely a calcium activated chloride channel. Mutations in the Ano5 gene have been associated with gnathodiaphyseal dysplasia. Alternatively spliced transcript variants have been described for Ano5. Diseases associated with ANO5 include Gnathodiaphyseal Dysplasia and Miyoshi Muscular Dystrophy 3.
          Bioinformatics
          Protein Aliases: Anoctamin-5; Gnathodiaphyseal dysplasia 1 protein; Gnathodiaphyseal dysplasia 1 protein homolog; integral membrane protein GDD1; Transmembrane protein 16E
          View more View less
          Gene Aliases: 9330162L24; ANO5; GDD1; LGMD2L; LGMDR12; TMEM16E
          View more View less
          UniProt ID: (Human) Q75V66, (Mouse) Q75UR0
          View more View less
          Entrez Gene ID: (Human) 203859, (Mouse) 233246
          View more View less
          Function(s)
          intracellular calcium activated chloride channel activity chloride channel activity protein dimerization activity phospholipid scramblase activity
          Process(es)
          chloride transport chloride transmembrane transport plasma membrane repair ion transmembrane transport
          France flag icon
          France
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline