          • Primary Antibodies ›
          • ANO5 Antibodies
          Antibodies Inc
          ANO5/TMEM16E Monoclonal Antibody (N421A/85)
          View all (9) ANO5 antibodies
          Tech Support
          Cite ANO5/TMEM16E Monoclonal Antibody (N421A/85)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          ANO5/TMEM16E Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          ANO5/TMEM16E Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          ANO5/TMEM16E Antibody (75-410) in WB
          Immunoblot against extracts of COS cells transiently transfected with untagged ANO5/TMEM16E, GFP- tagged ANO6/TMEM16F or untagged Kv2.1 plasmid probed with N421A/85 (left) or N86/38 (right) TC supe. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          ANO5/TMEM16E Antibody in Western Blot (WB)
          ANO5/TMEM16E Monoclonal Antibody (N421A/85)
          Product Details
          75-410
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000
          -
          Immunocytochemistry (ICC/IF)
          1:100
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N421A/85
          Immunogen
          Fusion protein amino acids 1-299 (cytoplasmic N-terminus) of human ANO5 (accession number Q75V66) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • FL490
          • FL550
          • FL594
          • FL650
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          TBS, pH 7.4
          Contains
          0.065% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          Ano5 does not exhibit calcium-activated chloride channel (CaCC) activity. The Ano5 gene encodes a member of the anoctamin family of transmembrane proteins. The Ano5 protein is likely a calcium activated chloride channel. Mutations in the Ano5 gene have been associated with gnathodiaphyseal dysplasia. Alternatively spliced transcript variants have been described for Ano5. Diseases associated with ANO5 include Gnathodiaphyseal Dysplasia and Miyoshi Muscular Dystrophy 3.
          Bioinformatics
          Protein Aliases: Anoctamin-5; Gnathodiaphyseal dysplasia 1 protein; Gnathodiaphyseal dysplasia 1 protein homolog; integral membrane protein GDD1; Transmembrane protein 16E
          View more View less
          Gene Aliases: 9330162L24; ANO5; GDD1; LGMD2L; LGMDR12; TMEM16E
          View more View less
          UniProt ID: (Human) Q75V66, (Mouse) Q75UR0
          View more View less
          Entrez Gene ID: (Human) 203859, (Mouse) 233246
          View more View less
          Function(s)
          intracellular calcium activated chloride channel activity chloride channel activity protein dimerization activity phospholipid scramblase activity
          Process(es)
          chloride transport chloride transmembrane transport plasma membrane repair ion transmembrane transport
          Contact tech support
          オンラインオーダー Plus Icon Minus Icon
          • オンラインオーダー可能な製品
          • オンラインオーダーに関する個人情報の取扱いについて
          • 特定商取引法に基づく表記
          • 返品について
          • 表示価格について
          • 配送料について
          サポート Plus Icon Minus Icon
          • FAQ、サポート情報
          • お問い合わせ一覧
          • サポートセンター
          • ドキュメント、証明書
          • 研究情報公開
          • Webサイトの問題を報告​
          リソース Plus Icon Minus Icon
          • ラーニングセンター
          • キャンペーン
          • イベント、セミナー
          • Blog
          • 企業情報
          • Social Responsibility (CSR)
          • お客さま体験の向上
          • 採用情報 採用情報
          • ニュース
          • 商標
          • ポリシーと通知
          • PPD
          • 標準販売条件
          • 個人情報保護方針
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline