SLC18A1 (Solute Carrier Family 18 Member A1) is a Protein Coding gene. Diseases associated with SLC18A1 include Barbiturate Abuse and Adrenal Neuroblastoma. Among its related pathways are Transport of inorganic cations/anions and amino acids/oligopeptides and Synaptic vesicle pathway. Gene Ontology (GO) annotations related to this gene include monoamine transmembrane transporter activity and xenobiotic transmembrane transporter activity. An important paralog of this gene is SLC18A2.
Protein Aliases: CGAT; solute carrier family 18 (vesicular monoamine transporter), member 1; solute carrier family 18 (vesicular monoamine), member 1; solute carrier family 18 member 1; solute carrier family18 (vesicular monoamine) member 1 (chromaffin granule amine transporter); unnamed protein product; VAT1; vesicular amine transporter 1; vesicular monoamine transporter 1; VMAT; VMAT 1; vesicular monoamine transporter 1; VMAT1
Gene Aliases: 4832416I10Rik; CGAT; Vat1; VMAT-1; Vmat1
UniProt ID: (Mouse) Q8R090, (Rat) Q01818
Entrez Gene ID: (Mouse) 110877, (Rat) 25693