    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TALK-1 Antibodies
          Antibodies Inc
          TALK1 Potassium Channel Monoclonal Antibody (N466/14), FL650
          View all (8) TALK-1 antibodies
          Tech Support
          Cite TALK1 Potassium Channel Monoclonal Antibody (N466/14), FL650
          Additional Information:
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          TALK1 Potassium Channel Monoclonal Antibody (N466/14), FL650
          Product Details
          75-461-FL650
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Mouse, Rat
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N466/14
          Immunogen
          Fusion protein amino acids 42-94 (first extracellular loop) and 259-292 (cytoplasmic C-terminus) of mouse TALK1 (accession number G5E845) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          FL650 FL650 FL650
          • Unconjugated
          • FL490
          • FL550
          • FL594
          • Request custom conjugation
          Excitation/Emission Max
          655/676 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          The KCNK16 gene encodes the TALK-1 channel, an abundant K+ channel transcript found in mouse and in human Beta-cells. There are four human TALK-1 transcript variants, including two that form functional K+ channels (TALK-1a (transcript variant 2) and TALK1-b (transcript variant 3). The TALK-1 structure includes 4 transmembrane domains, 2 extracellular loops, and intracellular N- and C-termini. The C-terminal tail of TALK-1 is a critical component that influences the magnitude of TALK-1 channel activation, while the N-terminus is important for protein interaction. TALK-1, two-pore domain K+ (K2p 16.1) channel is responsible for modulating glucose stimulated insulin secretion that relies on Beta-cell CA2+ influx. The channel produces outwardly rectifying, non-inactivating K+ currents, which are enhanced by elevations in extracellular pH. TALK-1 is thus pH-sensitive, with increased activity under alkaline conditions. In addition, it can be activated by singlet oxygen and nitric oxide.
          Bioinformatics
          Protein Aliases: 2P domain potassium channel Talk-1; potassium channel, subfamily K, member 16; potassium channel, two pore domain subfamily K, member 16; TWIK-related alkaline pH-activated K(+) channel 1
          View more View less
          Gene Aliases: 4731413G05Rik; TALK-1; TALK1
          View more View less
          UniProt ID: (Mouse) G5E845
          View more View less
          Entrez Gene ID: (Mouse) 74571, (Rat) 688996
          View more View less
          Function(s)
          potassium channel activity outward rectifier potassium channel activity potassium ion leak channel activity identical protein binding protein heterodimerization activity
          Process(es)
          potassium ion transport endoplasmic reticulum calcium ion homeostasis regulation of membrane potential regulation of release of sequestered calcium ion into cytosol regulation of insulin secretion involved in cellular response to glucose stimulus regulation of glucagon secretion potassium ion transmembrane transport membrane repolarization during action potential regulation of somatostatin secretion regulation of action potential
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-green-79565875f6-sf8lr:80/100.66.74.151:80.
          git-commit: 280dcbe45276d5d6e613c14d399020b8a49b7116
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.57.0-Offline