This gene encodes a conserved protein containing proline and serine rich regions. These regions may be important in protein-protein interactions.
Protein Aliases: Proline and serine-rich protein 1; unnamed protein product
Gene Aliases: 2810046L04Rik; 9330161F11; C13orf23; KIAA2032; PROSER1; RGD1309403
UniProt ID: (Human) Q86XN7, (Mouse) Q5PRE5
Entrez Gene ID: (Human) 80209, (Mouse) 212127, (Rat) 310417