TPTE encodes a putative transmembrane tyrosine phosphatase that may be involved in signal transduction pathways of the endocrine or spermatogenetic function of the testis.
Protein Aliases: Dr-VSP; voltage-sensing phosphoinositide phosphatase
Gene Aliases: tpip; vsp; wu:fd20e11; wu:fi24b06
Entrez Gene ID: (Zebrafish) 327292