    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • PTEN2 Antibodies
          Antibodies Inc
          VSP/TPTE Monoclonal Antibody (N432/63), FL550
          View all (18) PTEN2 antibodies
          Tech Support
          Cite VSP/TPTE Monoclonal Antibody (N432/63), FL550
          Additional Information:
          {{banner.description}}
          VSP/TPTE Monoclonal Antibody (N432/63), FL550
          Product Details
          75-486-FL550
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Chicken, Xenopus, Zebrafish
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N432/63
          Immunogen
          Fusion protein amino acids 153-335 of zebrafish TPTE (accession number Q4V9E4) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          FL550 FL550 FL550
          • Unconjugated
          • FL490
          • FL594
          • FL650
          • Request custom conjugation
          Excitation/Emission Max
          550/575 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          TPTE encodes a putative transmembrane tyrosine phosphatase that may be involved in signal transduction pathways of the endocrine or spermatogenetic function of the testis.
          Bioinformatics
          Protein Aliases: Dr-VSP; voltage-sensing phosphoinositide phosphatase
          View more View less
          Gene Aliases: tpip; vsp; wu:fd20e11; wu:fi24b06
          View more View less
          Entrez Gene ID: (Zebrafish) 327292
          View more View less
          Function(s)
          protein tyrosine phosphatase activity ion channel activity protein tyrosine/serine/threonine phosphatase activity phosphatidylinositol-3,4,5-trisphosphate 3-phosphatase activity protein phosphatase
          Process(es)
          protein dephosphorylation ion transport negative regulation of cell proliferation phosphatidylinositol 3-kinase signaling dephosphorylation phosphatidylinositol dephosphorylation cell motility regulation of protein kinase B signaling transmembrane transport
          • News News
          • Responsibility Responsibility
          México flag icon
          México
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline