    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
          • Order Status
          • 快速訂購
          • 登入
            登入
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • Vimentin Antibodies
          Invitrogen
          Vimentin Recombinant Mouse Monoclonal Antibody (V9)
          Advanced Verification
          View all (120) Vimentin antibodies
          Tech Support
          Cite Vimentin Recombinant Mouse Monoclonal Antibody (V9)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (4)
          • Advanced Verification (3)
          Vimentin  Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Vimentin  Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 7
          Vimentin Antibody (750013M) in ICC/IF
          Immunofluorescence analysis of Vimentin was performed using Vimentin Recombinant Mouse Monoclonal Antibody (V9) (Product # 750013M) on 70-80% confluent HeLa and MCF-7 cells. The cell lines were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton X-100 for 15 minutes and blocked with 2% BSA for 1 hour at room temperature. The cells were stained with Vimentin Recombinant Mouse Monocl... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Vimentin  Antibody in Immunocytochemistry (ICC/IF)
          Vimentin  Antibody in Western Blot (WB)
          Vimentin  Antibody in Flow Cytometry (Flow)
          Vimentin  Antibody in Flow Cytometry (Flow)
          Vimentin  Antibody
          Vimentin  Antibody
          Vimentin  Antibody
          Product Details
          750013M
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:400
          -
          Immunocytochemistry (ICC/IF)
          1:200
          -
          Flow Cytometry (Flow)
          0.03 µg/test
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1
          Expression System
          Expi293
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          V9
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • Alexa Fluor Plus 488 
          • Alexa Fluor Plus 555 
          • Alexa Fluor Plus 647 
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A/G
          Storage buffer
          PBS, pH 7.4
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Wet ice
          RRID
          AB_3719674
          Product Specific Information
          Recombinant mouse monoclonal antibodies are produced using in vitro expression systems. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include better specificity and lot-to-lot consistency.
          It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
          Target Information
          Vimentin is a developmentally regulated intermediate filament protein (IFP) found in cells of mesenchymal origin. It is believed to be involved with the intracellular transport of proteins between the nucleus and plasma membrane. Unlike other IFP proteins, vimentin is expressed, along with desmin, during the early stages of cellular development. During the development process, vimentin is exchanged for new, tissue-specific IFPs. Vimentin has been implicated to be involved in the rate of steroid synthesis via its role as a storage network for steroidogenic cholesterol containing lipid droplets. Vimentin phosphorylation by a protein kinase causes the breakdown of intermediate filaments and activation of an ATP and myosin light chain dependent contractile event. This results in cytoskeletal changes that facilitate the interaction of the lipid droplets within mitochondria, and subsequent transport of cholesterol to the organelles leading to an increase in steroid synthesis.
          Bioinformatics
          Protein Aliases: epididymis secretory sperm binding protein; FLJ36605; Hel113; RP11-124N14.1; unnamed protein product; VIM; VIME; Vimentin
          View more View less
          Gene Aliases: VIM
          View more View less
          UniProt ID: (Human) P08670
          View more View less
          Entrez Gene ID: (Human) 7431
          View more View less
          Function(s)
          RNA binding double-stranded RNA binding structural constituent of cytoskeleton structural constituent of eye lens protein binding protein domain specific binding identical protein binding binding, bridging scaffold protein binding keratin filament binding
          Process(es)
          positive regulation of gene expression negative regulation of neuron projection development astrocyte development neuron projection development positive regulation of collagen biosynthetic process regulation of mRNA stability intermediate filament-based process intermediate filament organization Bergmann glial cell differentiation lens fiber cell development cellular response to lipopolysaccharide cellular response to muramyl dipeptide cellular response to interferon-gamma
          Hong Kong flag icon
          Hong Kong
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline