Immunogen sequence: VARKFLDDEA ELSEEDAEYV SSDENDESEN EQDSSLLDFL NDETQLSQAI NDSEMRAIYM KSLRSPMMNN KYKMIHKTHK NINIFSQIPE QDETYLEDSF CVDEEESCKG QSSEEEVCVD FNLITDDCFA NSKKYKTRRA VMLKEMMEQN CAHSKKKLSR IILPDDSSEE ENNVNDKRES NIAVNPSTVK KNKQQDHCL
The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2). The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. This gene encodes the protein for complementation group M. Alternative splicing results in multiple transcript variants.
Protein Aliases: ATP-dependent RNA helicase FANCM; FAAP250; FANCM; Fanconi anemia complementation group M; Fanconi anemia group M protein; Fanconi anemia-associated polypeptide of 250 kDa; Protein FACM; Protein Hef ortholog; unnamed protein product
Gene Aliases: FAAP250; FANCM; KIAA1596; POF15; SPGF28
UniProt ID: (Human) Q8IYD8
Entrez Gene ID: (Human) 57697