    • 엔터프라이즈급 실험실 정보 처리
    • 360° CDMO 및 CRO 서비스
    • CDMO 서비스
    • 임상시험 CRO 서비스
    • 장비 서비스
    • 교육 서비스
    • Unity Lab Services
    • 서비스 전체 보기
  • 지원
    • 고객센터
    • 문의하기
    • 시험성적서(COA) 및 적합성 인증서(COC)
    • Safety Data Sheets (SDS)
    • 매뉴얼
    • 인용 및 참고 문헌
    • 기기 지원
    • 기술 자료/제품 FAQ
    • 학습 센터
    • 지원 메뉴 전체보기
      Cell Culture Plastics
  • 문의하기
  • 빠른 주문
  • 주문 현황
  • 제품 문서 검색
Thermo Fisher Scientific Logo
Search
전체검색
Search button
          • 주문 현황
          • 빠른주문
          • 로그인
            로그인
            회원이 아니신가요? 계정 생성하기
            • 계정정보
            • 주문내역조회
            • 커스텀제품 및 프로젝트
            • Services Central
          • Primary Antibodies ›
          • FANCM Antibodies
          Proteintech
          FANCM Recombinant Rabbit Monoclonal Antibody (230233G5), CoraLite® Plus 488
          View all (7) FANCM antibodies
          Tech Support
          Cite FANCM Recombinant Rabbit Monoclonal Antibody (230233G5), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          FANCM Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FANCM Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          FANCM Antibody (CL488-82897-2) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed Hela cells using CoraLite® Plus 488 FANCM antibody (CL488-82897-2, Clone: 230233G5) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          FANCM Antibody in Immunocytochemistry (ICC/IF)
          FANCM Antibody in Flow Cytometry (Flow)
          FANCM Recombinant Rabbit Monoclonal Antibody (230233G5), CoraLite® Plus 488
          Product Details
          CL488-82897-2
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Flow Cytometry (Flow)
          0.40 µg/10^6 cells in a 100 µL suspension
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          CHO cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230233G5
          Immunogen
          FANCM fusion protein Ag34447
          View immunogen
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: VARKFLDDEA ELSEEDAEYV SSDENDESEN EQDSSLLDFL NDETQLSQAI NDSEMRAIYM KSLRSPMMNN KYKMIHKTHK NINIFSQIPE QDETYLEDSF CVDEEESCKG QSSEEEVCVD FNLITDDCFA NSKKYKTRRA VMLKEMMEQN CAHSKKKLSR IILPDDSSEE ENNVNDKRES NIAVNPSTVK KNKQQDHCL
          Target Information
          The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2). The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. This gene encodes the protein for complementation group M. Alternative splicing results in multiple transcript variants.
          Bioinformatics
          Protein Aliases: ATP-dependent RNA helicase FANCM; FAAP250; FANCM; Fanconi anemia complementation group M; Fanconi anemia group M protein; Fanconi anemia-associated polypeptide of 250 kDa; Protein FACM; Protein Hef ortholog; unnamed protein product
          View more View less
          Gene Aliases: FAAP250; FANCM; KIAA1596; POF15; SPGF28
          View more View less
          UniProt ID: (Human) Q8IYD8
          View more View less
          Entrez Gene ID: (Human) 57697
          View more View less
          Function(s)
          nucleotide binding four-way junction DNA binding nucleic acid binding DNA binding chromatin binding RNA helicase activity helicase activity nuclease activity protein binding ATP binding four-way junction helicase activity hydrolase activity ATPase activity 3'-5' DNA helicase activity
          Process(es)
          resolution of meiotic recombination intermediates DNA repair cellular response to DNA damage stimulus replication fork processing reciprocal DNA recombination interstrand cross-link repair double-strand break repair via synthesis-dependent strand annealing positive regulation of protein monoubiquitination
          Contact tech support
          온라인 주문 Plus Icon Minus Icon
          • 주문 현황
          • 주문 지원
          • 빠른 주문
          • Supply Center
          • eProcurement
          지원 Plus Icon Minus Icon
          • Help and Support
          • 고객 센터
          • 기술 지원 센터
          • 제품 문서 검색
          • 사이트 문제 보고
          교육 및 이벤트 Plus Icon Minus Icon
          • 교육 센터
          • 프로모션
          • 이벤트 및 웨비나
          • 소셜 미디어
          • 소개 소개
          • 채용 채용
          • 투자자 투자자
          • 뉴스 뉴스
          • 사회적 책임 사회적 책임
          • Trademarks
          • 공정거래 공정거래
          • 정책 및 고지
          • PPD
          • 이용 약관
          • 개인정보 처리방침
          • 가격 및 운임 정책
          Korea flag icon
          Korea
          고객센터 문의 | 평일 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          장비 서비스 문의 | 평일 09:00~18:00
          1661-5055   |   Live Chat
          고객센터 문의 | 평일 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          장비 서비스 문의 | 평일 09:00~18:00
          1661-5055   |   Live Chat
          고객센터 문의 | 평일 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          장비 서비스 문의 | 평일 09:00~18:00
          1661-5055   |   Live Chat
          써모 피셔 사이언티픽 코리아 주식회사
          대표자 : 석수진
          사업자 등록번호 : 117-81-46910
          입금계좌 : 하나은행 336-890014-06204
          (예금주 : 써모피셔사이언티픽코리아 주식회사)
          써모 피셔 사이언티픽 솔루션스 유한회사
          대표자 : 석수진
          사업자 등록번호 : 114-86-04783
          입금계좌 : 신한은행 140-004-396660
          (예금주 : 써모피셔사이언티픽솔루션스 유한회사)
          주소 : 서울시 강남구 광평로 281 수서오피스빌딩 12층 06349 | 통신판매업신고번호 : 2015-서울강남-00898
          ISMS Logo
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline