    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • BAZ2A Antibodies
          Proteintech
          BAZ2A Recombinant Rabbit Monoclonal Antibody (230205B8)
          View all (10) BAZ2A antibodies
          Tech Support
          Cite BAZ2A Recombinant Rabbit Monoclonal Antibody (230205B8)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (5)
          BAZ2A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          BAZ2A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 5
          BAZ2A Antibody (82910-2-RR) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed U2OS cells using BAZ2A antibody (82910-2-RR, Clone: 230205B8) at dilution of 1:500 and CoraLite®488-Conjugated Goat Anti-Rabbit IgG (H+L) (SA00013-2). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          BAZ2A Antibody in Immunocytochemistry (ICC/IF)
          BAZ2A Antibody in Immunocytochemistry (ICC/IF)
          BAZ2A Antibody in Western Blot (WB)
          BAZ2A Antibody in Flow Cytometry (Flow)
          BAZ2A Antibody in Surface plasmon resonance (SPR)
          BAZ2A Recombinant Rabbit Monoclonal Antibody (230205B8)
          Product Details
          82910-2-RR
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:5,000-1:50,000
          -
          Immunocytochemistry (ICC/IF)
          1:250-1:1,000
          -
          Flow Cytometry (Flow)
          0.25 µg/10^6 cells in a 100 µL suspension
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Surface plasmon resonance (SPR)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          CHO cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230205B8
          Immunogen
          BAZ2A fusion protein Ag33963
          View immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: QYPSANPGSN LKDPPLLSQF SGGQYPLNGI LGGSRQPSSP SHNTNLRAGS QEFWANGTQS PMGLNFDSQE LYDSFPDQNF EVMPNGPPSF FTSPQTSPML GSSIQTFAPS QEVGSGIHPD EAAEKEMTSV VAENGTGLVG SLELEEEQPE LKMCGYNGSV PSVESLHQEV SVLVPDPTVS CLDDPSHLPD QLEDTPILS
          Target Information
          TIP 5 is the large subunit of the nucleolar remodeling complex NoRC. NoRC causes the repression of ribosomal gene transcription. It was demonstrated that histone deacetylation is involved in this repression and that TIP5 is associated with the histone deacetylase HDAC1 and mediates the deacetylation of histones in the vicinity of the rDNA promoter. The interaction of TIP5 and HDAC1, which is necessary for transcriptional repression, is established by the C-terminal PHD finger and bromodomain.
          Bioinformatics
          Protein Aliases: Bromodomain adjacent to zinc finger domain protein 2A; hWALp3; Transcription termination factor I-interacting protein 5; TTF-I interacting peptide 5; TTF-I-interacting protein 5; unnamed protein product
          View more View less
          Gene Aliases: BAZ2A; KIAA0314; TIP5; WALp3
          View more View less
          UniProt ID: (Human) Q9UIF9
          View more View less
          Entrez Gene ID: (Human) 11176
          View more View less
          Function(s)
          RNA polymerase I CORE element sequence-specific DNA binding DNA binding RNA binding protein binding zinc ion binding ligand-dependent nuclear receptor binding histone binding metal ion binding histone H4K16ac reader activity
          Process(es)
          chromatin silencing at rDNA RNA polymerase I preinitiation complex assembly chromatin organization chromatin remodeling methylation-dependent chromatin silencing transcription, DNA-templated regulation of transcription, DNA-templated negative regulation of transcription from RNA polymerase I promoter heterochromatin assembly
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline