    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • RORA Antibodies
          Proteintech
          RORA Recombinant Rabbit Monoclonal Antibody (230217G6)
          View all (21) RORA antibodies
          Tech Support
          Cite RORA Recombinant Rabbit Monoclonal Antibody (230217G6)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (8)
          RORA Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          RORA Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 8
          RORA Antibody (82930-1-RR) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed HepG2 cells using RORA antibody (82930-1-RR, Clone: 230217G6) at dilution of 1:300 and CoraLite®488-Conjugated AffiniPure Goat Anti-Rabbit IgG (H+L) (SA00013-2), CL594-Phalloidin (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          RORA Antibody in Immunocytochemistry (ICC/IF)
          RORA Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RORA Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RORA Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RORA Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RORA Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          RORA Antibody in Western Blot (WB)
          RORA Antibody in Flow Cytometry (Flow)
          RORA Recombinant Rabbit Monoclonal Antibody (230217G6)
          Product Details
          82930-1-RR
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:3,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:175-1:700
          -
          Immunohistochemistry (PFA fixed) (IHC (PFA))
          1:350
          -
          Immunocytochemistry (ICC/IF)
          1:150-1:600
          -
          Flow Cytometry (Flow)
          0.40 µg/10^6 cells in a 100 µL suspension
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          CHO cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230217G6
          Immunogen
          RORA fusion protein Ag0944
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          0.3 mg/mL
          Amount
          30 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: LDIQPSPDQS GLDINGIKPE PICDYTPASG FFPYCSFTNG ETSPTVSMAE LEHLAQNISK SHLETCQYLR EELQQITWQT FLQEEIENYQ NKQREVMWQL CAIKITEAIQ YVVEFAKRID GFMELCQNDQ IVLLKAGSLE VVFIRMCRAF DSQNNTVYFD GKYASPDVFK SLGCEDFISF VFEFGKSLCS MHLTEDEIAL FSAFVLMSAD RSWLQEKVKI EKLQQKIQLA LQHVLQKNHR EDGILTKLIC KVSTLRALCG RHTEKLMAFK AIYPDIVRLH FPPLYKELFT SEFEPAMQID G
          Target Information
          RORA is a receptor for retinoic acid belonging to the NR1 subfamily of nuclear hormone receptors with a nuclear receptor DNA binding domain. RORA binds either as a monomer or as a homodimer to the retinoic acid response element and thus regulates gene expression and also controls cell function. Though the exact function of RORA is yet unknown, it interacts with NM23-2, a nucleoside diphosphate kinase involved in organogenesis and differentiation, NM23-1, the product of a tumor metastasis suppressor candidate gene, CDK7, NCOA3 and NCOA6 coactivators. It is found to express in mesenchymal stem cells derived from bone marrow and are known to act by direct modulation of a bone matrix component. It is widely expressed in most of the tissues. Chromosomal aberrations involving RORA may be a cause of acute promyelocytic leukemia (APL).
          Bioinformatics
          Protein Aliases: Nuclear receptor ROR-alpha; Nuclear receptor RZR-alpha; Nuclear receptor subfamily 1 group F member 1; putative new variant; RAR-related orphan nuclear receptor alpha; RAR-related orphan receptor A; RAR-related orphan receptor alpha; retinoic acid receptor-related orphan receptor alpha; retinoid-related orphan receptor alpha; Retinoid-related orphan receptor-alpha; ROR Alpha; ROR-alpha; Roralpha; transcription factor RZR-alpha
          View more View less
          Gene Aliases: 9530021D13Rik; IDDECA; nmf267; NR1F1; ROR1; ROR2; ROR3; RORA; RORa1; RORalpha; RZR-ALPHA; RZRA; sg; staggerer; tmgc26
          View more View less
          UniProt ID: (Human) P35398, (Mouse) P51448
          View more View less
          Entrez Gene ID: (Human) 6095, (Rat) 300807, (Mouse) 19883
          View more View less
          Function(s)
          RNA polymerase II regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcription cofactor binding transcription corepressor binding transcription coactivator binding DNA binding transcription factor activity, sequence-specific DNA binding RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding protein binding beta-catenin binding oxysterol binding zinc ion binding sequence-specific DNA binding metal ion binding transcription factor activity, direct ligand regulated sequence-specific DNA binding core promoter sequence-specific DNA binding steroid hormone receptor activity transcription factor binding C4 zinc finger nuclear receptor
          Process(es)
          angiogenesis regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter xenobiotic metabolic process nitric oxide biosynthetic process circadian rhythm regulation of smoothened signaling pathway regulation of gene expression positive regulation of vascular endothelial growth factor production regulation of glucose metabolic process regulation of steroid metabolic process cerebellar Purkinje cell differentiation cerebellar granule cell precursor proliferation intracellular receptor signaling pathway circadian regulation of gene expression cellular response to sterol cholesterol homeostasis muscle cell differentiation regulation of circadian rhythm positive regulation of circadian rhythm regulation of macrophage activation negative regulation of I-kappaB kinase/NF-kappaB signaling negative regulation of fat cell differentiation positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter cGMP metabolic process rhythmic process negative regulation of inflammatory response triglyceride homeostasis cellular response to interleukin-1 cellular response to tumor necrosis factor cellular response to hypoxia T-helper 17 cell differentiation transcription, DNA-templated steroid hormone mediated signaling pathway regulation of transcription involved in cell fate commitment regulation of cholesterol homeostasis
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline