Immunogen sequence: LDIQPSPDQS GLDINGIKPE PICDYTPASG FFPYCSFTNG ETSPTVSMAE LEHLAQNISK SHLETCQYLR EELQQITWQT FLQEEIENYQ NKQREVMWQL CAIKITEAIQ YVVEFAKRID GFMELCQNDQ IVLLKAGSLE VVFIRMCRAF DSQNNTVYFD GKYASPDVFK SLGCEDFISF VFEFGKSLCS MHLTEDEIAL FSAFVLMSAD RSWLQEKVKI EKLQQKIQLA LQHVLQKNHR EDGILTKLIC KVSTLRALCG RHTEKLMAFK AIYPDIVRLH FPPLYKELFT SEFEPAMQID G
RORA is a receptor for retinoic acid belonging to the NR1 subfamily of nuclear hormone receptors with a nuclear receptor DNA binding domain. RORA binds either as a monomer or as a homodimer to the retinoic acid response element and thus regulates gene expression and also controls cell function. Though the exact function of RORA is yet unknown, it interacts with NM23-2, a nucleoside diphosphate kinase involved in organogenesis and differentiation, NM23-1, the product of a tumor metastasis suppressor candidate gene, CDK7, NCOA3 and NCOA6 coactivators. It is found to express in mesenchymal stem cells derived from bone marrow and are known to act by direct modulation of a bone matrix component. It is widely expressed in most of the tissues. Chromosomal aberrations involving RORA may be a cause of acute promyelocytic leukemia (APL).
Protein Aliases: Nuclear receptor ROR-alpha; Nuclear receptor RZR-alpha; Nuclear receptor subfamily 1 group F member 1; putative new variant; RAR-related orphan nuclear receptor alpha; RAR-related orphan receptor A; RAR-related orphan receptor alpha; retinoic acid receptor-related orphan receptor alpha; retinoid-related orphan receptor alpha; Retinoid-related orphan receptor-alpha; ROR Alpha; ROR-alpha; Roralpha; transcription factor RZR-alpha
Gene Aliases: IDDECA; NR1F1; ROR1; ROR2; ROR3; RORA; RORa1; RORalpha; RZR-ALPHA; RZRA
UniProt ID: (Human) P35398
Entrez Gene ID: (Human) 6095, (Rat) 300807