    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
          • Order Status
          • 快速訂購
          • 登入
            登入
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • Golgin-97 Antibodies
          Proteintech
          golgin 97 Recombinant Rabbit Monoclonal Antibody (230523A5)
          View all (20) Golgin-97 antibodies
          Tech Support
          Cite golgin 97 Recombinant Rabbit Monoclonal Antibody (230523A5)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (5)
          golgin 97 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          golgin 97 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 5
          golgin 97 Antibody (83009-4-RR) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed HepG2 cells using golgin 97 antibody (83009-4-RR, Clone: 230523A5) at dilution of 1:250 and CoraLite®488-Conjugated AffiniPure Goat Anti-Rabbit IgG (H+L) (SA00013-2), CL594-Phalloidin (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          golgin 97 Antibody in Immunocytochemistry (ICC/IF)
          golgin 97 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          golgin 97 Antibody in Western Blot (WB)
          golgin 97 Antibody in Flow Cytometry (Flow)
          golgin 97 Antibody in Surface plasmon resonance (SPR)
          golgin 97 Recombinant Rabbit Monoclonal Antibody (230523A5)
          Product Details
          83009-4-RR
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:5,000-1:20,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:50-1:500
          -
          Immunocytochemistry (ICC/IF)
          1:125-1:500
          -
          Flow Cytometry (Flow)
          0.25 µg/10^6 cells in a 100 µL suspension
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Surface plasmon resonance (SPR)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230523A5
          Immunogen
          golgin 97 fusion protein Ag3335
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: QIVALERMRA ADQTTAEQGM RQLEQENAAL KECRNEYERS LQNHQFELKK LKEEWSQREI VSVAMAQALE EVRKQREEFQ QQAANLTAII DEKEQNLREK TEVLLQKEQE ILQLERGHNS ALLQIHQLQA ELEALRTLKA EEAAVVAEQE DLLRLRGPLQ AEALSVNESH VTSRAMQDPV FQLPTAGRTP NGEVGAMDLT QLQKEKQDLE QQLLEKNKTI KQMQQRMLEL RKTLQKELKI RPDNELFEVR EKPGPEMANM APSVTNNTDL TDAREINFEY LKHVVLKFMS CRESEAFHLI KAVSVLLNFS QEEENMLKET LEYKMSWFGS KPAPKGSIRP SISNPRIPWS
          Target Information
          The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. The Golgins are a family of proteins, of which the protein encoded by this gene is a member, that are localized to the Golgi. This encoded protein is associated with Sjogren's syndrome.
          Bioinformatics
          Protein Aliases: gap junction protein, alpha 4, 37kD; golgi autoantigen, golgin subfamily a, 1; Golgin subfamily A member 1; Golgin-97; unnamed protein product
          View more View less
          Gene Aliases: GOLGA1; golgin-97
          View more View less
          UniProt ID: (Human) Q92805
          View more View less
          Entrez Gene ID: (Human) 2800
          View more View less
          Function(s)
          protein binding molecular carrier activity
          Process(es)
          cell-cell adhesion mediated by cadherin transepithelial transport
          Hong Kong flag icon
          Hong Kong
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline