    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
          • Primary Antibodies ›
          • Golgin-97 Antibodies
          Proteintech
          golgin 97 Recombinant Rabbit Monoclonal Antibody (230523A5), CoraLite® Plus 488
          View all (20) Golgin-97 antibodies
          Tech Support
          Cite golgin 97 Recombinant Rabbit Monoclonal Antibody (230523A5), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          golgin 97 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          golgin 97 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          golgin 97 Antibody (CL488-83009-4) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed HepG2 cells using CoraLite Plus 488 golgin 97 antibody (CL488-83009-4, Clone: 230523A5 ) at dilution of 1:200, CL594-Phalloidin (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          golgin 97 Antibody in Immunocytochemistry (ICC/IF)
          Product Details
          CL488-83009-4
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230523A5
          Immunogen
          golgin 97 fusion protein Ag3335
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: QIVALERMRA ADQTTAEQGM RQLEQENAAL KECRNEYERS LQNHQFELKK LKEEWSQREI VSVAMAQALE EVRKQREEFQ QQAANLTAII DEKEQNLREK TEVLLQKEQE ILQLERGHNS ALLQIHQLQA ELEALRTLKA EEAAVVAEQE DLLRLRGPLQ AEALSVNESH VTSRAMQDPV FQLPTAGRTP NGEVGAMDLT QLQKEKQDLE QQLLEKNKTI KQMQQRMLEL RKTLQKELKI RPDNELFEVR EKPGPEMANM APSVTNNTDL TDAREINFEY LKHVVLKFMS CRESEAFHLI KAVSVLLNFS QEEENMLKET LEYKMSWFGS KPAPKGSIRP SISNPRIPWS
          Target Information
          The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. The Golgins are a family of proteins, of which the protein encoded by this gene is a member, that are localized to the Golgi. This encoded protein is associated with Sjogren's syndrome.
          Bioinformatics
          Protein Aliases: gap junction protein, alpha 4, 37kD; golgi autoantigen, golgin subfamily a, 1; Golgin subfamily A member 1; Golgin-97; unnamed protein product
          View more View less
          Gene Aliases: GOLGA1; golgin-97
          View more View less
          UniProt ID: (Human) Q92805
          View more View less
          Entrez Gene ID: (Human) 2800
          View more View less
          Function(s)
          protein binding molecular carrier activity
          Process(es)
          cell-cell adhesion mediated by cadherin transepithelial transport
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          India flag icon
          India
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline