    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
  • Services
    • 360° CDMO and CRO Solutions
          • Primary Antibodies ›
          • HIC2 Antibodies
          Proteintech
          HIC2 Recombinant Rabbit Monoclonal Antibody (230177E8)
          View all (9) HIC2 antibodies
          Tech Support
          Cite HIC2 Recombinant Rabbit Monoclonal Antibody (230177E8)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (4)
          HIC2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          HIC2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          HIC2 Antibody (83162-7-RR) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed HepG2 cells using HIC2 antibody (83162-7-RR, Clone: 230177E8) at dilution of 1:400 and CoraLite®488-Conjugated Goat Anti-Rabbit IgG (H+L) (SA00013-2), CL594-Phalloidin (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          HIC2 Antibody in Immunocytochemistry (ICC/IF)
          HIC2 Antibody in Immunocytochemistry (ICC/IF)
          HIC2 Antibody in Flow Cytometry (Flow)
          HIC2 Antibody in Surface plasmon resonance (SPR)
          HIC2 Recombinant Rabbit Monoclonal Antibody (230177E8)
          Product Details
          83162-7-RR
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:200-1:800
          -
          Flow Cytometry (Flow)
          0.25 µg/10^6 cells in a 100 µL suspension
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Surface plasmon resonance (SPR)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230177E8
          Immunogen
          HIC2 fusion protein Ag18789
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: RSQRLSTASV IQARYQGLVD GRKGAHAPQE LPQAKGSDDE LFLGGSNQDS VQGLGRAVCP AGGEAGLGGC SSSTNGSSGG CEQELGLDLS KKSPPLPPAT PGPHLTPDDA AQLSDSQHGS PPAASAPPVA NSASYSELGG TPDEPMDLEG AEDNHLSLLE APGGQPRKSL RHSTRKKEWG KKEPVAGSPF ERREAGPKGP CPGEEGEGVG DRVPNGILAS GAGPSGPYGE PPYPCKEEEE NGKDASEDSA QSGSEGGSGH ASAHYMYRQE
          Target Information
          HIC2 contains 5 C2H2-type zinc fingers and 1 BTB (POZ) domain. It belongs to the krueppel C2H2-type zinc-finger protein family, Hic subfamily and is a transcriptional repressor.
          Bioinformatics
          Protein Aliases: Em:AP000557.1; Hic-2; Hic-3; HIC1-related gene on chromosome 22 protein; Hypermethylated in cancer 2 protein; Zinc finger and BTB domain-containing protein 30
          View more View less
          Gene Aliases: HIC2; HRG22; KIAA1020; ZBTB30; ZNF907
          View more View less
          UniProt ID: (Human) Q96JB3
          View more View less
          Entrez Gene ID: (Human) 23119
          View more View less
          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding protein binding zinc ion binding metal ion binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter regulation of cytokine production regulation of immune system process negative regulation of transcription, DNA-templated
          Hong Kong flag icon
          Hong Kong
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline