    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
          • Primary Antibodies ›
          • HIC2 Antibodies
          Proteintech
          HIC2 Recombinant Rabbit Monoclonal Antibody (230177E8), CoraLite® Plus 488
          View all (12) HIC2 antibodies
          Tech Support
          Cite HIC2 Recombinant Rabbit Monoclonal Antibody (230177E8), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          HIC2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          HIC2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          HIC2 Antibody (CL488-83162-7) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed HepG2 cells using CoraLite® Plus 488 HIC2 antibody (CL488-83162-7, Clone: 230177E8) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          HIC2 Antibody in Immunocytochemistry (ICC/IF)
          HIC2 Antibody in Flow Cytometry (Flow)
          HIC2 Recombinant Rabbit Monoclonal Antibody (230177E8), CoraLite® Plus 488
          Product Details
          CL488-83162-7
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Flow Cytometry (Flow)
          0.40 µg/10^6 cells in a 100 µL suspension
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230177E8
          Immunogen
          HIC2 fusion protein Ag18789
          3D Epitope / Immunogen
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: RSQRLSTASV IQARYQGLVD GRKGAHAPQE LPQAKGSDDE LFLGGSNQDS VQGLGRAVCP AGGEAGLGGC SSSTNGSSGG CEQELGLDLS KKSPPLPPAT PGPHLTPDDA AQLSDSQHGS PPAASAPPVA NSASYSELGG TPDEPMDLEG AEDNHLSLLE APGGQPRKSL RHSTRKKEWG KKEPVAGSPF ERREAGPKGP CPGEEGEGVG DRVPNGILAS GAGPSGPYGE PPYPCKEEEE NGKDASEDSA QSGSEGGSGH ASAHYMYRQE
          Target Information
          HIC2 contains 5 C2H2-type zinc fingers and 1 BTB (POZ) domain. It belongs to the krueppel C2H2-type zinc-finger protein family, Hic subfamily and is a transcriptional repressor.
          Bioinformatics
          Protein Aliases: Em:AP000557.1; Hic-2; Hic-3; HIC1-related gene on chromosome 22 protein; Hypermethylated in cancer 2 protein; Zinc finger and BTB domain-containing protein 30
          View more View less
          Gene Aliases: HIC2; HRG22; KIAA1020; ZBTB30; ZNF907
          View more View less
          UniProt ID: (Human) Q96JB3
          View more View less
          Entrez Gene ID: (Human) 23119
          View more View less
          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding protein binding zinc ion binding metal ion binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter regulation of cytokine production regulation of immune system process negative regulation of transcription, DNA-templated
          • Technical Support Centers
          • Find Documents and Certificates
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline