    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
          • Primary Antibodies ›
          • DOCK8 Antibodies
          Proteintech
          DOCK8 Recombinant Rabbit Monoclonal Antibody (230441A2)
          View all (27) DOCK8 antibodies
          Tech Support
          Cite DOCK8 Recombinant Rabbit Monoclonal Antibody (230441A2)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (5)
          DOCK8 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          DOCK8 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 5
          DOCK8 Antibody (83270-2-RR) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed THP-1 cells using DOCK8 antibody (83270-2-RR, Clone: 230441A2) at dilution of 1:200 and CoraLite®488-Conjugated AffiniPure Goat Anti-Rabbit IgG (H+L) (SA00013-2). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          DOCK8 Antibody in Immunocytochemistry (ICC/IF)
          DOCK8 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DOCK8 Antibody in Western Blot (WB)
          DOCK8 Antibody in Flow Cytometry (Flow)
          DOCK8 Antibody in Flow Cytometry (Flow)
          DOCK8 Recombinant Rabbit Monoclonal Antibody (230441A2)
          Product Details
          83270-2-RR
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:5,000-1:50,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:50-1:500
          -
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Flow Cytometry (Flow)
          0.25 µg/10^6 cells in a 100 µL suspension
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230441A2
          Immunogen
          DOCK8 fusion protein Ag34585
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: EVYKLVIPIL EAHREFRKLT LTHSKLQRAF DSIVNKDHKR MFGTYFRVGF FGSKFGDLDE QEFVYKEPAI TKLPEISHRL EAFYGQCFGA EFVEVIKDST PVDKTKLDPN KAYIQITFVE PYFDEYEMKD RVTYFEKNFN LRRFMYTTPF TLEGRPRGEL HEQYRRNTVL TTMHAFPYIK TRISVIQKEE FVLTPIEVAI EDMKKKTLQL AVAINQEPPD AKMLQMVLQG SVGATVNQGP LEVAQVFLAE IPADPKLYRH HNKLRLCFKE FIMRCGEAVE KNKRLITADQ REYQQELKKN YNKLKENLRP MIERKIPELY KPIFRVESQK RDSFHRSSFR KCETQLSQGS
          Target Information
          The Dedicator of cytokinesis protein 8 (DOCK8) is a member of the DOCK180 family of guanine nucleotide exchange factors. DOCK8 plays an essential role in humoral immune responses and is important in the proper formation of the B cell immunological synapse. Mutations in this gene result in the autosomal recessive form of the hyper-IgE syndrome.
          Bioinformatics
          Protein Aliases: 1200017A24Rik; Dedicator of cytokinesis protein 8; epididymis luminal protein 205; unnamed protein product
          View more View less
          Gene Aliases: DOCK8; HEL-205; HIES2; MRD2; ZIR8
          View more View less
          UniProt ID: (Human) Q8NF50
          View more View less
          Entrez Gene ID: (Human) 81704
          View more View less
          Function(s)
          guanyl-nucleotide exchange factor activity protein binding
          Process(es)
          immunological synapse formation small GTPase mediated signal transduction regulation of Rho protein signal transduction dendritic cell migration positive regulation of GTPase activity regulation of small GTPase mediated signal transduction memory T cell proliferation negative regulation of T cell apoptotic process positive regulation of establishment of T cell polarity cellular response to chemokine positive regulation of T cell migration
          • Technical Support Centers
          • Find Documents and Certificates
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline