    • See all product categories
  • Applications
    • Cell Analysis
    • Protein Biology
    • Lab Informatics
    • 360° CDMO and CRO Solutions
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
          • Primary Antibodies ›
          • TRMT6 Antibodies
          Proteintech
          TRMT6 Recombinant Rabbit Monoclonal Antibody (240911C5)
          View all (5) TRMT6 antibodies
          Tech Support
          Cite TRMT6 Recombinant Rabbit Monoclonal Antibody (240911C5)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (5)
          TRMT6 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          TRMT6 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 5
          TRMT6 Antibody (83843-5-RR) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed A431 cells using TRMT6 antibody (83843-5-RR, Clone: 240911C5) at dilution of 1:250 and CoraLite®488-Conjugated AffiniPure Goat Anti-Rabbit IgG (H+L) (SA00013-2), CL594-Phalloidin (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          TRMT6 Antibody in Immunocytochemistry (ICC/IF)
          TRMT6 Antibody in Immunocytochemistry (ICC/IF)
          TRMT6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TRMT6 Antibody in Western Blot (WB)
          TRMT6 Antibody in Surface plasmon resonance (SPR)
          TRMT6 Recombinant Rabbit Monoclonal Antibody (240911C5)
          Product Details
          83843-5-RR
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:5,000-1:50,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:1,500-1:6,000
          -
          Immunocytochemistry (ICC/IF)
          1:125-1:500
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Surface plasmon resonance (SPR)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          240911C5
          Immunogen
          TRMT6 fusion protein Ag10176
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MEGSGEQPGP QPQHPGDHRI RDGDFVVLKR EDVFKAVQVQ RRKKVTFEKQ WFYLDNVIGH SYGTAFEVTS GGSLQPKKKR EEPTAETKEA GTDNRNIVDD GKSQKLTQDD IKALKDKGIK GEEIVQQLIE NSTTFRDKTE FAQDKYIKKK KKKYEAIITV VKPSTRILSI MYYAREPGKI NHMRYDTLAQ MLTLGNIRAG NKMIVMETCA GLVLGAMMER MGGFGSIIQL YPGGGPVRAA TACFGFPKSF LSGLYEFPLN KVDSLLHGTF SAKMLSSEPK DSALVEESNG TLEEKQASEQ ENEDSMAEAP ESNHPEDQET METISQDPEH KGPKERGSKK DYIQEKQRRQ EEQRKRHLEA AALLSERNAD GLIVASRFHP TPLLLSLLDF VAPSRPFVVY CQYKEPLLEC YTKLRERGGV INLRLSETWL RNYQVLPDRS HPKLLMSGGG GYLLSGFTVA MDNLKADTSL KSNASTLESH ETEEPAAKKR KCPESDS
          Target Information
          Substrate-binding subunit of tRNA (adenine-N(1)-)-methyltransferase, which catalyzes the formation of N(1)-methyladenine at position 58 (m1A58) in initiator methionyl-tRNA.
          Bioinformatics
          Protein Aliases: MGC5029; mRNA methyladenosine-N(1)-methyltransferase non-catalytic subunit TRM6; tRNA (adenine(58)-N(1))-methyltransferase non-catalytic subunit TRM6; tRNA methyltransferase 6 homolog; tRNA(m1A58)-methyltransferase subunit TRM6; tRNA(m1A58)MTase subunit TRM6; unnamed protein product
          View more View less
          Gene Aliases: CGI-09; GCD10; Gcd10p; KIAA1153; TRM6; TRMT6
          View more View less
          UniProt ID: (Human) Q9UJA5
          View more View less
          Entrez Gene ID: (Human) 51605
          View more View less
          Function(s)
          RNA binding protein binding
          Process(es)
          mRNA processing tRNA processing tRNA methylation
          • Trademarks
          • Fair Trade Fair Trade
          Korea flag icon
          Korea
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910
          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783
          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)
          ISMS Logo
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline