    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
          • Primary Antibodies ›
          • TRMT6 Antibodies
          Proteintech
          TRMT6 Recombinant Rabbit Monoclonal Antibody (240911C5), CoraLite® Plus 488
          View all (7) TRMT6 antibodies
          Tech Support
          Cite TRMT6 Recombinant Rabbit Monoclonal Antibody (240911C5), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          TRMT6 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          TRMT6 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          TRMT6 Antibody (CL488-83843-5) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed HeLa cells using CoraLite Plus 488 TRMT6 antibody (CL488-83843-5, Clone: 240911C5 ) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          TRMT6 Antibody in Immunocytochemistry (ICC/IF)
          TRMT6 Recombinant Rabbit Monoclonal Antibody (240911C5), CoraLite® Plus 488
          Product Details
          CL488-83843-5
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          240911C5
          Immunogen
          TRMT6 fusion protein Ag10176
          View immunogen
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: MEGSGEQPGP QPQHPGDHRI RDGDFVVLKR EDVFKAVQVQ RRKKVTFEKQ WFYLDNVIGH SYGTAFEVTS GGSLQPKKKR EEPTAETKEA GTDNRNIVDD GKSQKLTQDD IKALKDKGIK GEEIVQQLIE NSTTFRDKTE FAQDKYIKKK KKKYEAIITV VKPSTRILSI MYYAREPGKI NHMRYDTLAQ MLTLGNIRAG NKMIVMETCA GLVLGAMMER MGGFGSIIQL YPGGGPVRAA TACFGFPKSF LSGLYEFPLN KVDSLLHGTF SAKMLSSEPK DSALVEESNG TLEEKQASEQ ENEDSMAEAP ESNHPEDQET METISQDPEH KGPKERGSKK DYIQEKQRRQ EEQRKRHLEA AALLSERNAD GLIVASRFHP TPLLLSLLDF VAPSRPFVVY CQYKEPLLEC YTKLRERGGV INLRLSETWL RNYQVLPDRS HPKLLMSGGG GYLLSGFTVA MDNLKADTSL KSNASTLESH ETEEPAAKKR KCPESDS
          Target Information
          Substrate-binding subunit of tRNA (adenine-N(1)-)-methyltransferase, which catalyzes the formation of N(1)-methyladenine at position 58 (m1A58) in initiator methionyl-tRNA.
          Bioinformatics
          Protein Aliases: MGC5029; mRNA methyladenosine-N(1)-methyltransferase non-catalytic subunit TRM6; tRNA (adenine(58)-N(1))-methyltransferase non-catalytic subunit TRM6; tRNA methyltransferase 6 homolog; tRNA(m1A58)-methyltransferase subunit TRM6; tRNA(m1A58)MTase subunit TRM6; unnamed protein product
          View more View less
          Gene Aliases: CGI-09; GCD10; Gcd10p; KIAA1153; TRM6; TRMT6
          View more View less
          UniProt ID: (Human) Q9UJA5
          View more View less
          Entrez Gene ID: (Human) 51605
          View more View less
          Function(s)
          RNA binding protein binding
          Process(es)
          mRNA processing tRNA processing tRNA methylation
          • Technical Support Centers
          • Find Documents and Certificates
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline