    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PEG3 Antibodies
          Proteintech
          PEG3 Recombinant Rabbit Monoclonal Antibody (241234C5)
          View all (16) PEG3 antibodies
          Tech Support
          Cite PEG3 Recombinant Rabbit Monoclonal Antibody (241234C5)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (7)
          PEG3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PEG3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 7
          PEG3 Antibody (84030-5-RR) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed HepG2 cells using PEG3 antibody (84030-5-RR, Clone: 241234C5) at dilution of 1:300 and CoraLite®488-Conjugated AffiniPure Goat Anti-Rabbit IgG (H+L) (SA00013-2), CL594-Phalloidin (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          PEG3 Antibody in Immunocytochemistry (ICC/IF)
          PEG3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PEG3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PEG3 Antibody in Western Blot (WB)
          PEG3 Antibody in Western Blot (WB)
          PEG3 Antibody in Flow Cytometry (Flow)
          PEG3 Antibody in Surface plasmon resonance (SPR)
          Product Details
          84030-5-RR
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:2,000-1:10,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:200-1:800
          -
          Immunocytochemistry (ICC/IF)
          1:150-1:600
          -
          Flow Cytometry (Flow)
          0.25 µg/10^6 cells in a 100 µL suspension
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Surface plasmon resonance (SPR)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          241234C5
          Immunogen
          PEG3 fusion protein Ag20132
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: LNEHMRLHR EDDLLEQSQM AEEAIIPGLA LTEFQRSQTE ERLFECAVCG ESFVNPAELA DHVTVHKNEP YEYGSSYTHT SFLTEPLKGA IPFYECKDCG KSFIHSTVLT KHKELHLEEE EEDEAAAAAA AAAQEVEANV HVPQVVLRIQ GLNVEAAEPE VEAAEPEVEA AEPEVEAAEP NGEAEGPDGE AAEPIGEAGQ PNGEAEQPNG DADEPDGAGI EDPEERAEEP EGKAEEPEGD ADEPDGVGIE DPEEGEDQEI QVEEPYYDCH ECTETFTSST AFSEHLKTHA SMIIFEPANA FGECSGYIER ASTSTGGANQ ADEKYFKCDV CGQLFNDRLS LARHQNTHTG
          Target Information
          In human, ZIM2 and PEG3 are treated as two distinct genes though they share multiple 5' exons and a common promoter and both genes are paternally expressed. Alternative splicing events connect their shared 5' exons either with the remaining 4 exons unique to ZIM2, or with the remaining 2 exons unique to PEG3. In contrast, in other mammals ZIM2 does not undergo imprinting and, in mouse, cow, and likely other mammals as well, the ZIM2 and PEG3 genes do not share exons. Human PEG3 protein belongs to the Kruppel C2H2-type zinc finger protein family. PEG3 may play a role in cell proliferation and p53-mediated apoptosis. PEG3 has also shown tumor suppressor activity and tumorigenesis in glioma and ovarian cells. Alternative splicing of this PEG3 gene results in multiple transcript variants encoding distinct isoforms.
          Bioinformatics
          Protein Aliases: KIAA0287; Kruppel-type zinc finger protein; maternally imprinted; Paternally-expressed gene 3 protein; unnamed protein product; Zinc finger and SCAN domain-containing protein 24
          View more View less
          Gene Aliases: KIAA0287; PEG3; PW1; ZKSCAN22; ZNF904; ZSCAN24
          View more View less
          UniProt ID: (Human) Q9GZU2
          View more View less
          Entrez Gene ID: (Human) 5178
          View more View less
          Function(s)
          RNA polymerase II regulatory region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding zinc ion binding metal ion binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter regulation of transcription from RNA polymerase II promoter apoptotic process regulation of gene expression positive regulation of transcription from RNA polymerase II promoter
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline