    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • PEG3 Antibodies
          Proteintech
          PEG3 Recombinant Rabbit Monoclonal Antibody (241234C5), CoraLite® Plus 488
          View all (13) PEG3 antibodies
          Tech Support
          Cite PEG3 Recombinant Rabbit Monoclonal Antibody (241234C5), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          PEG3 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          PEG3 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          PEG3 Antibody (CL488-84030-5) in Flow
          1x10^6 HepG2 cells were intracellularly stained with 0.25 µg Coralite Plus 488 Peg3 Recombinant Antibody (CL488-84030-5, Clone: 241234C5) (red), or 0.25 µg CoraLite Plus 488 Rabbit IgG Isotype Control Recombinant Antibody (CL488-98136, Clone: 240953C9) (blue). Cells were fixed and permeabilized with Transcription Factor Staining Buffer Kit (PF,00011). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          PEG3 Antibody in Flow Cytometry (Flow)
          PEG3 Recombinant Rabbit Monoclonal Antibody (241234C5), CoraLite® Plus 488
          Product Details
          CL488-84030-5
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          0.25 µg per 10^6 cells
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          241234C5
          Immunogen
          PEG3 fusion protein Ag20132
          3D Epitope / Immunogen
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: LNEHMRLHRE DDLLEQSQMA EEAIIPGLAL TEFQRSQTEE RLFECAVCGE SFVNPAELAD HVTVHKNEPY EYGSSYTHTS FLTEPLKGAI PFYECKDCGK SFIHSTVLTK HKELHLEEEE EDEAAAAAAA AAQEVEANVH VPQVVLRIQG LNVEAAEPEV EAAEPEVEAA EPEVEAAEPN GEAEGPDGEA AEPIGEAGQP NGEAEQPNGD ADEPDGAGIE DPEERAEEPE GKAEEPEGDA DEPDGVGIED PEEGEDQEIQ VEEPYYDCHE CTETFTSSTA FSEHLKTHAS MIIFEPANAF GECSGYIERA STSTGGANQA DEKYFKCDVC GQLFNDRLSL ARHQNTHTG
          Target Information
          In human, ZIM2 and PEG3 are treated as two distinct genes though they share multiple 5' exons and a common promoter and both genes are paternally expressed. Alternative splicing events connect their shared 5' exons either with the remaining 4 exons unique to ZIM2, or with the remaining 2 exons unique to PEG3. In contrast, in other mammals ZIM2 does not undergo imprinting and, in mouse, cow, and likely other mammals as well, the ZIM2 and PEG3 genes do not share exons. Human PEG3 protein belongs to the Kruppel C2H2-type zinc finger protein family. PEG3 may play a role in cell proliferation and p53-mediated apoptosis. PEG3 has also shown tumor suppressor activity and tumorigenesis in glioma and ovarian cells. Alternative splicing of this PEG3 gene results in multiple transcript variants encoding distinct isoforms.
          Bioinformatics
          Protein Aliases: KIAA0287; Kruppel-type zinc finger protein; maternally imprinted; Paternally-expressed gene 3 protein; unnamed protein product; Zinc finger and SCAN domain-containing protein 24
          View more View less
          Gene Aliases: KIAA0287; PEG3; PW1; ZKSCAN22; ZNF904; ZSCAN24
          View more View less
          UniProt ID: (Human) Q9GZU2
          View more View less
          Entrez Gene ID: (Human) 5178
          View more View less
          Function(s)
          RNA polymerase II regulatory region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding zinc ion binding metal ion binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter regulation of transcription from RNA polymerase II promoter apoptotic process regulation of gene expression positive regulation of transcription from RNA polymerase II promoter
          • News News
          • Responsibility Responsibility
          Chile flag icon
          Chile
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline