  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • MTMR4 Antibodies
          Proteintech
          MTMR4 Recombinant Rabbit Monoclonal Antibody (241304E11)
          View all (4) MTMR4 antibodies
          Tech Support
          Cite MTMR4 Recombinant Rabbit Monoclonal Antibody (241304E11)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          MTMR4 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          MTMR4 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          MTMR4 Antibody (84328-1-RR) in ICC/IF
          Immunofluorescent analysis of (-20°C Ethanol) fixed HepG2 cells using MTMR4 antibody (84328-1-RR, Clone: 241304E11) at dilution of 1:250 and CoraLite®488-Conjugated Goat Anti-Rabbit IgG (H+L) (SA00013-2). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          MTMR4 Antibody in Immunocytochemistry (ICC/IF)
          MTMR4 Antibody in Surface plasmon resonance (SPR)
          Product Details
          84328-1-RR
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:125-1:500
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Surface plasmon resonance (SPR)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          241304E11
          Immunogen
          MTMR4 fusion protein Ag4310
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: SHQEQLSSV PDLTHGEEDI GKRGNNRNGQ LLENPRFGKM PLELVRKPIS QSQISEFSFL GSNWDSFQGM VTSFPSGEAT PRRLLSYGCC SKRPNSKQMR ATGPCFGGQW AQREGVKSPV CSSHSNGHCT GPGGKNQMWL SSHPKQVSST KPVPLNCPSP VPPLYLDDDG LPFPTDVIQH RLRQIEAGYK QEVEQLRRQV RELQMRLDIR HCCAPPAEPP MDYEDDFTCL KESDGSDTED FGSDHSEDCL SEASWEPVDK KETEVTRWVP DHMASHCYNC DCEFWLAKRR HHCRNCGNVF CAGCCHLKLP IPDQQLYDPV LVCNSCYEHI QVSRARELMS QQLKKPIATA SS
          Target Information
          MTMR, a member of the myotubularin family, encodes an intracellular protein of 1, 195 amino acids, and shares approximately 47% sequence identity with MTMR3, spiking to over 70% identity in the catalytic domain and the double zinc finger (FYVE) motif. PCR analysis detects expression of MTMR4 in all human tissues examined except lung, small intestine, stomach, salivary gland, adrenal gland, and uterus. Subcellular localization of MTMR4 in transfected HeLa cells showed a perinuclear distribution, similar to that seen with MTMR3. MTMR4 dephosphorylates para-nitrophenylphosphate and phosphatidylinositol 3-phosphate.
          Bioinformatics
          Protein Aliases: DKFZp781E194; FYVE domain-containing dual specificity protein phosphatase 2; FYVE-DSP2; MGC163449; MGC163451; MTMR7; Myotubularin-related protein 4; Phosphatidylinositol-3,5-bisphosphate 3-phosphatase; Phosphatidylinositol-3,5-bisphosphate 3-phosphatase MTMR4; Zinc finger FYVE domain-containing protein 11; zinc finger, FYVE domain containing 11
          View more View less
          Gene Aliases: FYVE-DSP2; KIAA0647; MTMR4; ZFYVE11
          View more View less
          UniProt ID: (Human) Q9NYA4
          View more View less
          Entrez Gene ID: (Human) 9110
          View more View less
          Function(s)
          phosphatidylinositol-3-phosphatase activity protein serine/threonine phosphatase activity protein tyrosine phosphatase activity protein binding zinc ion binding hydrolase activity protein phosphatase binding metal ion binding phosphatidylinositol-3,5-bisphosphate 3-phosphatase activity binding, bridging R-SMAD binding
          Process(es)
          phosphatidylinositol biosynthetic process negative regulation of transforming growth factor beta receptor signaling pathway negative regulation of BMP signaling pathway phosphatidylinositol dephosphorylation midbody abscission phagosome maturation negative regulation of endocytic recycling
          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline