    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
          • Primary Antibodies ›
          • 6x His Tag Antibodies
          GenScript
          His Tag Monoclonal Antibody (6G2A9), iFluor™ 555, THE™
          View all (109) 6x His Tag antibodies
          Tech Support
          Cite His Tag Monoclonal Antibody (6G2A9), iFluor™ 555, THE™
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          His Tag Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          His Tag Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          His Tag Antibody (A01801-100) in ICC/IF
          Immunocytochemistry analysis of non-transfected CHO cells (a) and His-tagged protein transfected CHO cells (b) using THE™ His Tag Antibody [iFluor 555], mAb, Mouse (A01801-100, 4 µg/mL). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          His Tag Antibody in Immunocytochemistry (ICC/IF)
          Product Details
          A01801-100
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1-4 µg/mL
          -
          Product Specifications
          Species Reactivity
          Tag
          Host/Isotype
          Mouse / IgG1, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          6G2A9
          Immunogen
          A synthetic peptide HHHHHH coupled to KLH.
          Conjugate
          iFluor™ 555 iFluor™ 555 iFluor™ 555
          • Unconjugated
          • Biotin 
          • FITC
          • HRP
          • iFluor 488
          • iFluor 647
          • Request custom conjugation
          Excitation/Emission Max
          557/570 nm
          Form
          Lyophilized
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.4, with 10mg/mL BSA
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Reconstitute the lyophilized product with deionized water (or equivalent) to make antibody concentration of 0.5 mg/mL.
          The His tag is a short amino acid sequence consists of Histidine (His) residues appended to recombinant proteins. Ni-charged resins are used for the purification of His-tagged proteins from eukaryotic and prokaryotic expression systems. An anti-His tag antibody is a useful tool for the analysis of His-tagged proteins. THE™ His Tag Antibody [iFluor 555], mAb, Mouse is THE™ His Tag Antibody (A00186) conjugated with iFluor 555 under optimal conditions with an F/P ratio of 3-6. It is suitable for detecting the expression level of His-tagged proteins. iFluor 555 is a bright and photostable fluorescent dye. It is an excellent alternative to Alexa Fluor 555.
          Upon reconstitution, it can be stored for 2-3 weeks at 2-8 °C or for up to 12 months at -20 °C or below.
          Target Information
          The 6x His tag is a synthetic oligo peptide consisting of 6 consecutive histidine residues (HHHHHH). The His tag is commonly expressed as a tag at either N-or C-terminal regions of recombinant proteins to allow isolation or purification by immobilized metal affinity chromatography. Epitope tagging is a technique in which a known epitope is fused to a recombinant protein using genetic engineering. By choosing a particular epitope and recombinant protein combination, epitope tagging makes it possible to detect proteins for which no antibody is available. His epitope tagged proteins that contain a stretch of histidine residues at the carboxyl or amino terminus are used for purification of recombinant proteins by means of metal chelate chromatography due to metal binding properties of histidine clusters on His tagged proteins. For example, the affinity of the histidine-tag motif to Ni2+ by chelation is strong and selective enough to enable purification of the protein to homogeneity by affinity chromatography on a Ni2+-NTA adsorbent. His tag-specific antibodies are used to facilitate detection or coimmunopreciptation of tagged proteins.
          Bioinformatics
          Protein Aliases: 6His tag; 6X His; His Tag; Histidine Tag
          • Technical Support Centers
          • Find Documents and Certificates
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline