    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • HA Tag Antibodies
          GenScript
          HA Tag Monoclonal Antibody (5E11D8), iFluor™ 647, THE™
          View all (94) HA Tag antibodies
          Tech Support
          Cite HA Tag Monoclonal Antibody (5E11D8), iFluor™ 647, THE™
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          HA Tag Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          HA Tag Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          HA Tag Antibody (A01808-100) in Flow
          Flow cytometric analysis of non-transfected CHO cells (Black) or HA-tagged protein transfected CHO cells (Red) using THE™ HA Tag Antibody [iFluor 647], mAb, Mouse (A01808-100, 4 µg/mL). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          HA Tag Antibody in Flow Cytometry (Flow)
          Product Details
          A01808-100
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1-4 µg/mL
          -
          Flow Cytometry (Flow)
          1-4 µg/mL
          -
          Product Specifications
          Species Reactivity
          Tag
          Host/Isotype
          Mouse / IgG1, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          5E11D8
          Immunogen
          A synthetic peptide YPYDVPDYA coupled to KLH
          Conjugate
          iFluor™ 647 iFluor™ 647 iFluor™ 647
          • Unconjugated
          • FITC
          • HRP
          • iFluor 488
          • Request custom conjugation
          Excitation/Emission Max
          656/670 nm
          Form
          Lyophilized
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.4, with 10mg/mL BSA
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Reconstitute the lyophilized product with deionized water (or equivalent) to make antibody concentration of 0.5 mg/mL.
          The HA tag is a polypeptide with the amino acid sequence YPYDVPDYA that can be added to a target protein using recombinant DNA technology. It can be fused to the N-terminus or C-terminus of the protein to facilitate detection and purification. An anti-HA tag antibody is a useful tool for the analysis of HA-tagged proteins.THE™ HA Tag Antibody [iFluor 647], mAb, Mouse is THE™ HA Tag Antibody (A01244) conjugated with iFluor 647 under optimal conditions with an F/P ratio of 3-6. It is suitable for detecting the expression level of HA-tagged proteins. iFluor 647 is a bright and photostable fluorescent dye. It is an excellent alternative to Alexa Fluor 647.
          Upon reconstitution, it can be stored for 2-3 weeks at 2-8 °C or for up to 12 months at -20 °C or below.
          Target Information
          HA Tag (hemagglutinin tag) is an epitope tag that provides a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. HA (hemagglutinin) is a surface glycoprotein derived from influenza hemagglutinin, and is commonly expressed as a tag in conjunction with proteins to aid in identification and purification of the tagged protein for functional analysis. HA tag antibodies can be used to facilitate protein detection or co-immunopreciptation of tagged proteins.
          Bioinformatics
          Protein Aliases: HA; HA Epitope Tag; HA Tag; HA-tag; hemagglutinin
          Argentina flag icon
          Argentina
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline