  • Applications
    • Bioprocédés
    • Culture cellulaire et transfection
    • Thérapie cellulaire et génétique
    • Chromatographie
    • Tests moléculaires
    • Solutions numériques
    • Extraction et analyse d’ADN et d’ARN
    • Spectroscopie, analyse élémentaire et isotopique
    • Voir toutes les applications et techniques
  • Services
    • Services CDMO et CRO à 360°
    • Services CDMO
    • Services CRO
    • Services personnalisés
    • Services financiers et leasing
    • Services pour les instruments
    • Informatique de laboratoire
    • Offres commerciales et OEM
    • Services de formation
    • Unity Lab Services
    • Voir tous les services
  • Aide et assistance
    • S’inscrire et créer un compte
    • Comment commander
    • Assistance instruments
    • Centres d’assistance technique
    • Centres d’apprentissage
    • Consultez toutes les rubriques d'aide et d'assistance
  • Populaire
    • TaqMan Real-Time PCR Assays
      Reaction PCR en temps réel TaqMan
    • Antibodies
      Anticorps
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt - Synthèse Gène
    • Cell Culture Plastics
      Plastiques culture cellulaire
  • Nos clients
    • Biotechnologie
    • Industrie biopharmaceutique
    • CDMO
    • Diagnostics de laboratoire
    • Sciences industrielles et appliquées associées
  • Promotions
  • Contactez-nous
  • Commande rapide
  • Suivi et statut de la commande
  • Documents et certificats
Thermo Fisher Scientific Logo
Search
Rechercher
Search button
          • Statut des commandes
          • Commande rapide
          • Promos
          • Se connecter
            Se connecter
            Vous n’avez pas de compte? Créer votre compte
            • Mon compte
            • Commandes
            • Connect: lab, données, apps
            • Produits personnalisés et projets
            • Services Central
          • Primary Antibodies ›
          • HSPE1 Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          Bethyl Laboratories
          HSPE1/CPN10/HSP10 Polyclonal Antibody
          View all (23) HSPE1 antibodies
          Tech Support
          Cite HSPE1/CPN10/HSP10 Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          HSPE1/CPN10/HSP10 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          HSPE1/CPN10/HSP10 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          HSPE1/CPN10/HSP10 Antibody (A304-842A-T) in WB
          Detection of human and mouse HSPE1/CPN10 by western blot. Samples: Whole cell lysate (50 µg) from HeLa, 293T, Jurkat, mouse TCMK-1, and mouse NIH3T3 cells prepared using NETN lysis buffer. Antibody: Affinity purified rabbit anti-HSPE1/CPN10 antibody A304-842A-M (lot A304-842A-M-1) used for WB at 1:1000. Detection: Chemiluminescence with an exposure time of 30 seconds. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          HSPE1/CPN10/HSP10 Antibody in Western Blot (WB)
          Product Details
          A304-842A-T
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000
          -
          Product Specifications
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Between 52 and 102
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          40 µg/mL
          Amount
          0.8 µg
          Storage conditions
          4°C
          Shipping conditions
          Wet ice
          Product Specific Information
          The recommended shelf life for this product is 1 year from date of receipt.
          Based on 100% sequence identity, this antibody is predicted to react with Rat and Bovine
          Target Information
          The heat shock proteins (HSPs) comprise a group of highly conserved, abundantly expressed proteins with diverse functions, including the assembly and sequestering of multiprotein complexes, transportation of nascent polypeptide chains across cellular membranes and regulation of protein folding. Heat shock proteins (also known as molecular chaperones) fall into six general families: HSP 90, HSP 70, HSP 60, the low molecular weight HSPs, the immunophilins and the HSP 110 family. The low molecular weight family includes HSP 10, HSP 20, HSP 27 (Heme Oxygenase 1), HSP 32 and HSP 40. Chaperonins are ubiquitous, indispensable proteins that facilitate protein folding in an ATP-dependent manner, enhancing the yield of properly folded substrate protein under conditions where spontaneous folding does not occur. Chaperonins are typified by the E. coli heat-shock proteins GroEL and GroES (HSP 10). HSP 10 is a heptameric ring of identical 10 kDa subunits that binds to each end of GroEL to form a symmetric, functional heterodimer.
          Contact tech support
          Commander Plus Icon Minus Icon
          • Statut des commandes
          • Assistance commandes
          • Commande rapide
          • Centre d’approvisionnement
          • Approvisionnement en ligne
          Assistance Plus Icon Minus Icon
          • Aide et assistance
          • Contactez-nous
          • Centres d’assistance technique
          • Documents et certificats
          • Signaler un problème sur le site
          Ressources Plus Icon Minus Icon
          • Centres d’apprentissage
          • Promotions
          • Événements et séminaires en ligne
          • Réseaux sociaux
          À propos de Thermo Fisher Plus Icon Minus Icon
          • À propos de nous À propos de nous
          • Emplois Emplois
          • Investisseurs Investisseurs
          • Actualités Actualités
          • Responsabilité sociale Responsabilité sociale
          • Marques commerciales
          • Politiques et notices d’information
          Notre portfolio Plus Icon Minus Icon
          France flag icon
          France
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline