          • Primary Antibodies ›
          • Frizzled-2 Antibodies
          Alomone Labs, LTD
          Frizzled-2 (FZD2) (extracellular) Polyclonal Antibody
          View all (10) Frizzled-2 antibodies
          Tech Support
          Cite Frizzled-2 (FZD2) (extracellular) Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (3)
          Frizzled-2 (FZD2) (extracellular) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Frizzled-2 (FZD2) (extracellular) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          Frizzled-2 (FZD2) (extracellular) Antibody (AFR-022-25UL) in WB
          Western blot analysis of human MDA-231 breast carcinoma cell line lysate (lanes 1 and 3) and human THP-1 acute monocytic leukemia cell line lysate (lanes 2 and 4): - 1,2. Anti-Frizzled-2 (FZD2) (extracellular) Antibody (#AFR-022), (1:200).3,4. Anti-Frizzled-2 (FZD2) (extracellular) Antibody , preincubated with Frizzled-2/FZD2 (extracellular) Blocking Peptide (#BLP-FR022). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Frizzled-2 (FZD2) (extracellular) Antibody in Western Blot (WB)
          Frizzled-2 (FZD2) (extracellular) Antibody in Western Blot (WB)
          Frizzled-2 (FZD2) (extracellular) Antibody in Flow Cytometry (Flow)
          Frizzled-2 (FZD2) (extracellular) Polyclonal Antibody
          Product Details
          AFR-022-25UL
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:200
          -
          Flow Cytometry (Flow)
          2.5 µg
          -
          Product Specifications
          Species Reactivity
          Human, Rat
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          (C)GAPPRYATLEHPFH, corresponding to amino acid residues 192-205 of rat Frizzled-2, Extracellular, N-terminus
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Lyophilized
          Concentration
          0.8 mg/mL
          Amount
          20 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          PBS, pH 7.4, with 1% BSA
          Contains
          0.05% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
          Target Information
          This intronless gene is a member of the frizzled gene family. Members of this family encode seven-transmembrane domain proteins that are receptors for the wingless type MMTV integration site family of signaling proteins. This gene encodes a protein that is coupled to the beta-catenin canonical signaling pathway. Competition between the wingless-type MMTV integration site family, member 3A and wingless-type MMTV integration site family, member 5A gene products for binding of this protein is thought to regulate the beta-catenin-dependent and -independent pathways.
          Bioinformatics
          Protein Aliases: Dfrizzled2; Drosophila polarity gene (frizzled) homologue; Drosophila polarity gene homolog 2; frizzled 2, seven transmembrane spanning receptor; frizzled family receptor 2; frizzled homolog 2; Frizzled-2; Fz-2; FzE2; polarity gene; putative; rFz2
          View more View less
          Gene Aliases: Fz-10; fz-2; Fz2; Fzd10; FZD2; fzE2; hFz2; OMOD2
          View more View less
          UniProt ID: (Human) Q14332, (Rat) Q08464
          View more View less
          Entrez Gene ID: (Human) 2535, (Rat) 64512
          View more View less
          Function(s)
          transmembrane signaling receptor activity G-protein coupled receptor activity protein binding Wnt-protein binding PDZ domain binding Wnt-activated receptor activity identical protein binding protein heterodimerization activity transmembrane signal receptor
          Process(es)
          membranous septum morphogenesis muscular septum morphogenesis outflow tract morphogenesis signal transduction cell surface receptor signaling pathway G-protein coupled receptor signaling pathway cell-cell signaling sensory perception of smell Wnt signaling pathway neuron differentiation non-canonical Wnt signaling pathway endothelial cell differentiation positive regulation of transcription, DNA-templated hard palate development canonical Wnt signaling pathway Wnt signaling pathway, planar cell polarity pathway inner ear receptor cell development ventricular septum morphogenesis cochlea morphogenesis cell activation G-protein coupled receptor signaling pathway coupled to cGMP nucleotide second messenger positive regulation of cytosolic calcium ion concentration Wnt signaling pathway, calcium modulating pathway multicellular organism development positive regulation of cGMP metabolic process epithelial cell differentiation positive regulation of sequence-specific DNA binding transcription factor activity cellular response to vitamin D cellular response to growth factor stimulus cellular response to peptide hormone stimulus planar cell polarity pathway involved in neural tube closure
          Pedidos Plus Icon Minus Icon
          • Status do pedido
          • Ajuda para pedidos
          • Pedido rápido
          • Supply Center
          • eProcurement
          Suporte Plus Icon Minus Icon
          • Ajuda e suporte
          • Entre em Contato
          • Centros de Suporte Técnico
          • Obter Documentos e Certificados
          • Informe um Problema no Site
          Recursos Plus Icon Minus Icon
          • Centros de aprendizagem
          • Promoções
          • Eventos & Webinars
          • Mídia Sociais
          Sobre a Thermo Fisher Plus Icon Minus Icon
          • Sobre Nós Sobre Nós
          • Carreiras Carreiras
          • Investidores Investidores
          • Sala de Impresa Sala de Impresa
          • Responsabilidade Social Responsabilidade Social
          • Marcas
          • Políticas e avisos
          Nosso Portfólio Plus Icon Minus Icon
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline