Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • AMIGO3 Antibodies

          Proteintech

          AMIGO3 Polyclonal Antibody

          View all (6) AMIGO3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite AMIGO3 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (7)
          AMIGO3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          AMIGO3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 7

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          AMIGO3 Antibody (21278-1-AP) in ICC/IF

          Immunofluorescent analysis of ( 4% PFA ) fixed MCF-7 cells using 21278-1-AP (AMIGO3 antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          AMIGO3 Antibody in Immunocytochemistry (ICC/IF)
          AMIGO3 Antibody in Western Blot (WB)
          AMIGO3 Antibody in Western Blot (WB)
          AMIGO3 Antibody in Western Blot (WB)
          AMIGO3 Antibody in Western Blot (WB)
          AMIGO3 Antibody in Western Blot (WB)
          AMIGO3 Antibody in Western Blot (WB)
          AMIGO3 Polyclonal Antibody

          Product Details

          21278-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:3,000
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          AMIGO3 Fusion Protein Ag15790 (221-504 aa encoded by BC110418)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.33 mg/mL

          Amount

          49.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: LPCDCRLYH LLQRWHQRGL SAVRDFAREY VCLAFKVPAS RVRFFQHSRV FENCSSAPAL GLERPEEHLY ALVGRSLRLY CNTSVPAMRI AWVSPQQELL RAPGSRDGSI AVLADGSLAI GNVQEQHAGL FVCLATGPRL HHNQTHEYNV SVHFPRPEPE AFNTGFTTLL GCAVGLVLVL LYLFAPPCRC CRRACRCRRW PQTPSPLQEL SAQSSVLSTT PPDAPSRKAS VHKHVVFLEP GRRGLNGRVQ LAVAEEFDLY NPGGLQLKAG SESASSIGSE GPMTT (221-504 aa encoded by BC110418)

          Target Information

          Amphoterin induced gene and ORF 3 (AMIGO3) is a gene located on chromosome 3p21.31, encoding a member of the AMIGO family of proteins involved in cell adhesion, axon guidance, and neuronal signaling. AMIGO3 is primarily expressed in the central nervous system, playing a crucial role in neural development and the maintenance of neural circuits. Functionally, AMIGO3 contributes to neuronal growth and synapse formation, impacting processes such as neural plasticity and signal transduction. The protein's activity facilitates interactions with other neuronal surface proteins, influencing axon pathfinding and connectivity. Aberrant expression or mutations in AMIGO3 have been linked to neurological disorders and neurodegenerative diseases, suggesting its significance in neural health and pathology. Research into AMIGO3 continues to explore its potential therapeutic applications in promoting neural repair and modulating synaptic functions, offering insights into strategies for addressing nervous system disorders.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: alivin 3; Alivin-3; AMIGO-3; amigo3; amigo3 {ECO:0000312|EMBL:AAO48952.1, ECO:0000312|RGD:631413}; amphoterin induced gene and ORF 3; amphoterin-induced gene and ORF 3; Amphoterin-induced protein 3; MGC120552; transmembrane protein AMIGO3

          View more View less

          Gene Aliases: ALI3; AMIGO-3; AMIGO3; E430002N15Rik; KIAA1851; mKIAA1851; UNQ6084/PRO20089

          View more View less

          UniProt ID: (Human) Q86WK7, (Rat) Q80ZD5, (Mouse) Q8C2S7

          View more View less

          Entrez Gene ID: (Human) 386724, (Rat) 316003, (Mouse) 320844

          View more View less

          Function(s)
          macromolecular complex binding protein binding molecular_function
          Process(es)
          heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules nervous system development brain development cell adhesion positive regulation of synapse assembly cell-cell adhesion negative regulation of neuron projection development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline