    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • BrdU Antibodies
          Invitrogen
          BrdU Monoclonal Antibody (MoBU-1)
          5 Published Figures
          13 References
          View all (98) BrdU antibodies
          Tech Support
          Cite BrdU Monoclonal Antibody (MoBU-1)
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Published Figures (5)
          BrdU Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          BrdU Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 5
          BrdU Antibody (B35141) in ICC/IF
          Development (Cambridge, England) 2022 - Fig. 5. HH signaling regulates HBE cell proliferation. (A) Timeline for EdU, BrdU and cyclopamine administration. (B) Immunostaining for Ki67 (violet), BrdU (green) and EdU fluorescence (red) in HBE cells at the ALI after ethanol ( n =6) or 10 muM cyclopamine ( n =6) treatment at day 3. (C) Immunostaining for Ki67 (violet), BrdU (green) and EdU fluorescence (red) in HBE cells at the ALI after ethanol ( n =6... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          BrdU Antibody in Immunocytochemistry (ICC/IF)
          BrdU Antibody in Flow Cytometry (Flow)
          BrdU Antibody in Flow Cytometry (Flow)
          BrdU Antibody in Flow Cytometry (Flow)
          BrdU Antibody in Flow Cytometry (Flow)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          Product Details
          B35141
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (IHC)
          -
          View 1 publication 1 publication
          Immunohistochemistry - Free Floating (IHC (Free))
          -
          View 1 publication 1 publication
          Immunocytochemistry (ICC/IF)
          -
          View 3 publications 3 publications
          Flow Cytometry (Flow)
          Assay-dependent
          View 7 publications 7 publications
          Miscellaneous PubMed (Misc)
          -
          View 1 publication 1 publication
          Product Specifications
          Species Reactivity
          Chemical
          Published species
          Chemical, Mouse
          Host/Isotype
          Mouse / IgG
          Class
          Monoclonal
          Type
          Antibody
          Clone
          MoBU-1
          Immunogen
          BrdU
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • Alexa Fluor 488 
          • Alexa Fluor 594 
          • Alexa Fluor 647 
          • Biotin 
          • Pacific Blue
          • Request custom conjugation
          Form
          Liquid
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.2
          Contains
          5mM sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2536441
          Target Information
          Bromodeoxyuridine (5-bromo-2-deoxyuridine, BrdU) is a synthetic nucleoside that is an analogue of thymidine. BrdU is commonly used in the detection of proliferating cells in living tissues, and can be incorporated into the newly synthesized DNA of replicating cells (during the S phase of the cell cycle), substituting for thymidine during DNA replication. Antibodies specific for BrdU can then be used to detect the incorporated chemical, thus indicating cells that were actively replicating their DNA. Binding of the antibody requires denaturation of the DNA, usually by exposing the cells to acid or heat.
          Bioinformatics
          Protein Aliases: 5-Bromo-2-deoxyuridine; Bromodeoxyuridine
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-green-7bf6cff7cf-v8ns5:80/100.66.75.228:80.
          git-commit: cd21911f92a61daaa7859949453b3286702c44a2
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/3.7.0-2026.11.47-1.0