    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • MUNC18 Antibodies
          Proteintech
          STXBP1 Monoclonal Antibody (1B5B3), Biotin
          View all (27) MUNC18 antibodies
          Tech Support
          Cite STXBP1 Monoclonal Antibody (1B5B3), Biotin
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          STXBP1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          STXBP1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          STXBP1 Antibody (BIOTIN-67137) in IHC (P)
          Immunohistochemical analysis of paraffin-embedded mouse brain tissue slide using Biotin-67137 (STXBP1 antibody) at dilution of 1:200 (under 40x lens). Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          STXBP1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          STXBP1 Monoclonal Antibody (1B5B3), Biotin
          Product Details
          BIOTIN-67137
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Paraffin) (IHC (P))
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Pig, Rat
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          1B5B3
          Immunogen
          STXBP1 fusion protein Ag28605
          3D Epitope / Immunogen
          Conjugate
          Biotin Biotin Biotin
          • Unconjugated
          • CoraLite 594
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1000 µg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: HELTFQAMSY DLLPIENDVY KYETSGIGEA RVKEVLLDED DDLWIALRHK HIAEVSQEVT RSLKDFSSSK RMNTGEKTTM RDLSQMLKKM PQYQKELSKY STHLHLAEDC MKHYQGTVDK LCRVEQDLAM GTDAEGEKIK DPMRAIVPIL LDANVSTYDK IRIILLYIFL KNGITEENLN KLIQHAQIPP EDSEIITNMA HLGVPIVTDS TLRRRSKPER KERISEQTYQ LSRWTPIIKD IMEDTIEDKL DTKHYPYIST RSSASFSTTA VSARYGHWHK NKAPGEYRSG PRLIIFILGG VSLNEMRCAY EVTQANGKWE VLIGSTHILT PQKLLDTLKK LNKTDEEISS
          Target Information
          This gene encodes a syntaxin-binding protein. The encoded protein appears to play a role in release of neurotransmitters via regulation of syntaxin, a transmembrane attachment protein receptor. Mutations in this gene have been associated with infantile epileptic encephalopathy-4. Alternatively spliced transcript variants have been described.
          Bioinformatics
          Protein Aliases: EIEE4; FLJ37475; hUNC18; MGC146482; minisatellite 10g detected by probe MMS10; munc-18c; MUNC18-1; N-Sec1; neuronal SEC1; p67; Protein unc-18 homolog 1; Protein unc-18 homolog A; rbSec1; rop; RP11-56D16.3; Syntaxin-binding protein 1; UNC-18 homolog; unc-18-1; Unc-18A; Unc18-1; unnamed protein product
          View more View less
          Gene Aliases: AI317162; AI326233; ANC18HA; DEE4; MMS10-G; Ms10g; Munc-18a; MUNC18-1; N-Sec1; NSEC1; NSEC1A; P67; Rb-sec1; RBSEC1; rbSec1A; rbSec1B; Sec1; STXBP1; Sxtbp1; unc-18A; UNC18; Unc18-1; UNC18A; Unc18h
          View more View less
          UniProt ID: (Human) P61764, (Mouse) O08599, (Rat) P61765
          View more View less
          Entrez Gene ID: (Pig) 100517927, (Human) 6812, (Mouse) 20910, (Rat) 25558
          View more View less
          Function(s)
          SNARE binding RNA binding protein binding syntaxin-1 binding protein kinase binding protein domain specific binding syntaxin binding identical protein binding phospholipase binding poly(A) RNA binding protein N-terminus binding membrane trafficking regulatory protein
          Process(es)
          platelet degranulation developmental process involved in reproduction intracellular protein transport exocytosis vesicle docking involved in exocytosis neurotransmitter secretion neuromuscular synaptic transmission axon target recognition regulation of synaptic vesicle priming protein transport synaptic vesicle priming synaptic vesicle maturation vesicle-mediated transport negative regulation of protein complex assembly regulation of vesicle fusion regulation of synaptic vesicle fusion to presynaptic membrane negative regulation of synaptic transmission, GABAergic response to estradiol SNARE complex assembly regulation of SNARE complex assembly positive regulation of mast cell degranulation negative regulation of neuron apoptotic process regulated exocytosis positive regulation of exocytosis positive regulation of calcium ion-dependent exocytosis protein stabilization neuron apoptotic process establishment of localization in cell long term synaptic depression platelet aggregation cellular response to interferon-gamma protein localization to plasma membrane presynaptic dense core vesicle exocytosis positive regulation of vesicle docking positive regulation of glutamate secretion, neurotransmission regulation of regulated secretory pathway regulation of acrosomal vesicle exocytosis transport synaptic vesicle fusion to presynaptic active zone membrane
          France flag icon
          France
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline