Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: CKRVEQKEEC VKLAETEETD KSETMETEDV PTSSRLYWAD LKTLLSEKLN SIEFADTIKQ LSQNTYTPRE AGSQKDESLA YYIENQFHEF KFSKVWRDEH YVKIQVKSSI GQNMVTIVQS NGNLDPVESP EGYVAFSKPT EVSGKLVHAN FGTKKDFEEL SYSVNGSLVI VRAGEITFAE KVANAQSFNA IGVLIYMDKN KFPVVEADLA LFGHAHLGTG DPYTPGFPSF NHTQFPPSQS SGLPNIPVQT ISRAAAEKLF GKMEGSCPAR WNIDSSCKLE LSQNQNVKLI VKNVLKERRI LNIFGVIKGY EEPDRYVVVG AQRDALGAGV AAKSSVGTGL LLKLAQVFSD MISKDGFRPS RSIIFASWTA GDFGAVGATE WLEGYLSSLH LKAFTYINLD KVVLGTSNFK VSASPLLYTL MGKIMQDVKH PVDGKSLYRD SNWISKVEKL SFDNAAYPFL AYSGIPAVSF CFCEDADYPY LGTRLDTYEA LTQKVPQLNQ MVRTAAEVAG QLIIKLTHDV ELNLDYEMYN SKLLSFMKDL NQFKTDIRDM GLSLQWLYSA RGDYFRATSR LTTDFHNAEK TNRFVMREIN DRIMKVEYHF LSPYVSPRES PFRHIFWGSG SHTLSALVEN LKLRQKNITA FNETLFRNQL ALATWTIQGV ANALSGDIWN IDNEF
Cellular uptake of iron occurs via receptor-mediated endocytosis of ligand-occupied transferrin receptor into specialized endosomes. Endosomal acidification leads to iron release. The apotransferrin-receptor complex is then recycled to the cell surface with a return to neutral pH and the concomitant loss of affinity of apotransferrin for its receptor. Transferrin receptor is necessary for development of erythrocytes and the nervous system. Serum transferrin receptor (sTfR) is used as a means of detecting erythropoietin (EPO) misuse by athletes and as a diagnostic test for anemias resulting from a number of conditions including rheumatoid arthritis, pregnancy, irritable bowel syndrome and in HIV patients.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD antigen CD71; CD71; mammary tumor virus receptor 1; p90; T9; TR; transferrin receptor; Transferrin receptor 1; Transferrin receptor protein 1; unnamed protein product
Gene Aliases: 2610028K12Rik; AI195355; AI426448; AU015758; CD71; E430033M20Rik; IMD46; Mtvr-1; Mtvr1; p90; T9; TFR; TFR1; TFRC; TR; TRFR
UniProt ID: (Human) Q1HE24, (Mouse) Q62351
Entrez Gene ID: (Human) 7037, (Mouse) 22042
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support