Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • Transferrin Receptor (CD71) Antibodies

          Invitrogen

          CD71 (Transferrin Receptor) Monoclonal Antibody (R17217 (RI7 217.1.4)), NovaFluor™ Blue 610-70S, eBioscience™

          View all (135) Transferrin Receptor (CD71) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CD71 (Transferrin Receptor) Monoclonal Antibody (R17217 (RI7 217.1.4)), NovaFluor™ Blue 610-70S, eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          CD71 (Transferrin Receptor) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD71 (Transferrin Receptor) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD71 (Transferrin Receptor) Antibody (M040T03B06-A) in Flow

          C57BL/6 mouse bone marrow cells were unstained (left) or stained with 0.05 µg of CD71 (Transferrin Receptor) Monoclonal Antibody, NovaFluor Blue 610-70S (right). All cells were co-stained with CD45R (B220) Monoclonal Antibody, eFluor 450 (Product # 48-0452-82). Total viable cells in the lymphocyte gate were used for analysis, as determined by LIVE/DEAD Blue (Product # L34962). Data was acquired on a 5-laser... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD71 (Transferrin Receptor) Antibody in Flow Cytometry (Flow)
          CD71 (Transferrin Receptor) Antibody in Flow Cytometry (Flow)

          Product Details

          M040T03B06-A

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          0.05 µg/test
          -
          Product Specifications

          Species Reactivity

          Mouse

          Host/Isotype

          Rat / IgG2a, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          R17217 (RI7 217.1.4)

          Conjugate

          NovaFluor™ Blue 610-70S NovaFluor™ Blue 610-70S NovaFluor™ Blue 610-70S
          • Unconjugated
          • Alexa Fluor 700 
          • APC 
          • APC-eFluor 780
          • Biotin 
          • eFluor 450
          • FITC
          • NovaFluor Blue 660-120S
          • NovaFluor Blue 690
          • NovaFluor Blue 800
          • NovaFluor Red 685
          • NovaFluor Red 710
          • NovaFluor Red 755
          • NovaFluor UV 765
          • NovaFluor Violet 690
          • NovaFluor Yellow 690
          • NovaFluor Yellow 730
          • NovaFluor Yellow 755
          • NovaFluor Yellow 810
          • PE 
          • PE-Cyanine7
          • PerCP-eFluor 710
          • Super Bright 436
          • Super Bright 600
          • Super Bright 645
          • Request custom conjugation

          Excitation/Emission Max

          492/616 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2, with BSA

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          RRID

          AB_3679431

          Product Specific Information

          Description
          The R17217 monoclonal antibody reacts with mouse CD71, a 170-180 kDa type II transmembrane protein. CD71, the transferrin receptor, exists as a homodimer on the cell surface and is essential for cellular growth. CD71 is expressed by immature proliferating cells and at low levels on resting mature lymphocytes. Antigen or mitogen stimulation of T and B cells upregulates the expression of CD71. Expression level differences have been observed in different mouse strains.

          This product contains 1 vial of NovaFluor conjugate and 1 vial of CellBlox Plus Blocking Buffer.

          Applications Tested
          This R17217 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.05 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Master mixes
          • Master mixes of NFs should be made at 2-8 °C and may be made up to 4 hours ahead of time.
          • We do not recommend storing master mixes containing NovaFluor conjugates overnight or longer.

          Whole Blood compatibility
          • When utilizing whole blood (as opposed to density-gradient-purified PBMC), we recommend lysing red blood cells in bulk prior to staining with NovaFluor conjugates.
          • See the Bulk Lysis of Human Whole Blood protocol here.
          • Staining of whole blood with NovaFluor conjugates followed by lysis of red blood cells may result in higher-than-expected background staining.

          Viability dye compatibility
          • NovaFluor dyes are not compatible with DNA intercalating viability dyes.
          • Do not use viability dyes such as propidium iodide, 7-actinomycin D (7-AAD) and DAPI. Invitrogen LIVE/DEAD Fixable Dead Cell stains are recommended for use with NovaFluor dyes.

          CellBlox Plus Blocking Buffer
          • This NovaFluor conjugate comes with CellBlox Plus Blocking Buffer (for 25 µg or 25 test products, Cat. No. C001T02F01; for 100 µg or 100 test, Cat. No. C001T03F01), essential for optimal staining.
          • Use CellBlox Plus Blocking Buffer in all experiments with NovaFluor conjugates.
          • Add 5 µL per sample to antibody cocktails/master mixes (regardless of how many Novafluor-conjugated antibodies are present) before combining with cells.
          • CellBlox Plus Blocking Buffer is compatible with either Super Bright Complete Blocking Buffer or Brilliant Stain Buffer and can be used in antibody cocktails/master mixes with those reagents.
          • For single-color controls, use 5 µL of CellBlox Plus Blocking Buffer per 100 µL of cell sample (10^3 to 10^8 cells).

          NovaFluor conjugates are based on Phiton technology utilizing novel fluorophore-containing nucleic acid dye structures that allow for engineered fluorescent signatures with consideration for spillover and spread impacts. Learn more

          Excitation: 492 nm; Emission: 616 nm; Laser: 488 nm (Blue) Laser

          Target Information

          Cellular uptake of iron occurs via receptor-mediated endocytosis of ligand-occupied transferrin receptor into specialized endosomes. Endosomal acidification leads to iron release. The apotransferrin-receptor complex is then recycled to the cell surface with a return to neutral pH and the concomitant loss of affinity of apotransferrin for its receptor. Transferrin receptor is necessary for development of erythrocytes and the nervous system. Serum transferrin receptor (sTfR) is used as a means of detecting erythropoietin (EPO) misuse by athletes and as a diagnostic test for anemias resulting from a number of conditions including rheumatoid arthritis, pregnancy, irritable bowel syndrome and in HIV patients.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CD antigen CD71; CD71; mammary tumor virus receptor 1; TR; Transferrin receptor protein 1

          View more View less

          Gene Aliases: 2610028K12Rik; AI195355; AI426448; AU015758; CD71; E430033M20Rik; Mtvr-1; Mtvr1; p90; TFR; TFR1; Tfrc; TR; Trfr

          View more View less

          UniProt ID: (Mouse) Q62351

          View more View less

          Entrez Gene ID: (Mouse) 22042

          View more View less

          Function(s)
          glycoprotein binding double-stranded RNA binding transferrin receptor activity iron ion transmembrane transporter activity protein binding Hsp70 protein binding transferrin transmembrane transporter activity identical protein binding poly(A) RNA binding chaperone binding metalloprotease
          Process(es)
          cellular iron ion homeostasis endocytosis receptor-mediated endocytosis osteoclast differentiation transferrin transport iron ion transmembrane transport cellular response to drug positive regulation of bone resorption protein transmembrane transport iron ion import
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline