FIGURE: 1 / 1
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
This gene encodes a member of the alpha tubulin protein family. Alpha tubulins are one of two core protein families (alpha and beta tubulins) that heterodimerize and assemble to form microtubules. Mutations in this gene are associated with polymicrogyria and optic nerve hypoplasia. Alternate splicing results in multiple transcript variants.
Protein Aliases: Alpha-tubulin 8; TUBA8; tubulin alpha 8 trucated form; Tubulin alpha chain-like 2; Tubulin alpha-8 chain; unnamed protein product
Gene Aliases: CDCBM8; MACTHC2; TUBA8; TUBAL2
UniProt ID: (Human) Q9NY65
Entrez Gene ID: (Human) 51807