          • Primary Antibodies ›
          • TUBA8 Antibodies
          OriGene
          TUBA8 Monoclonal Antibody (OTI3G2), Biotin, TrueMAB™
          View all (22) TUBA8 antibodies
          Tech Support
          Cite TUBA8 Monoclonal Antibody (OTI3G2), Biotin, TrueMAB™
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          TUBA8 Antibody in Luminex (Luminex)
          Group 53 Created with Sketch.
          TUBA8 Antibody in Luminex (Luminex)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          TUBA8 Antibody (TA700529) in Luminex
          TUBA8 Luminex with 2G6 Capture (TA600519) and 3G2 Detection (TA700529) Antibodies. Substrate used: Recombinant Human TUBA8 (TP311175) {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          TUBA8 Antibody in Luminex (Luminex)
          TUBA8 Monoclonal Antibody (OTI3G2), Biotin, TrueMAB™
          Product Details
          TA700529
          Applications
          Tested Dilution
          Publications
          ELISA (ELISA)
          1:250-1:1,000
          -
          Luminex (Luminex)
          Assay-Dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          OTI3G2
          Immunogen
          Full length human recombinant protein of human TUBA8 produced in HEK293T cell.
          3D Epitope / Immunogen
          Conjugate
          Biotin Biotin Biotin
          • Unconjugated
          • Request custom conjugation
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          50 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS with 10mg/mL BSA, 50% glycerol
          Contains
          0.05% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          This gene encodes a member of the alpha tubulin protein family. Alpha tubulins are one of two core protein families (alpha and beta tubulins) that heterodimerize and assemble to form microtubules. Mutations in this gene are associated with polymicrogyria and optic nerve hypoplasia. Alternate splicing results in multiple transcript variants.
          Bioinformatics
          Protein Aliases: Alpha-tubulin 8; TUBA8; tubulin alpha 8 trucated form; Tubulin alpha chain-like 2; Tubulin alpha-8 chain; unnamed protein product
          View more View less
          Gene Aliases: CDCBM8; MACTHC2; TUBA8; TUBAL2
          View more View less
          UniProt ID: (Human) Q9NY65
          View more View less
          Entrez Gene ID: (Human) 51807
          View more View less
          Function(s)
          nucleotide binding structural constituent of cytoskeleton protein binding GTP binding hydrolase activity metal ion binding
          Process(es)
          microtubule cytoskeleton organization mitotic cell cycle microtubule-based process spermatogenesis spermatid development
          Pedidos Plus Icon Minus Icon
          • Status do pedido
          • Ajuda para pedidos
          • Pedido rápido
          • Supply Center
          • eProcurement
          Suporte Plus Icon Minus Icon
          • Ajuda e suporte
          • Entre em Contato
          • Centros de Suporte Técnico
          • Obter Documentos e Certificados
          • Informe um Problema no Site
          Recursos Plus Icon Minus Icon
          • Centros de aprendizagem
          • Promoções
          • Eventos & Webinars
          • Mídia Sociais
          Sobre a Thermo Fisher Plus Icon Minus Icon
          • Sobre Nós Sobre Nós
          • Carreiras Carreiras
          • Investidores Investidores
          • Sala de Impresa Sala de Impresa
          • Responsabilidade Social Responsabilidade Social
          • Marcas
          • Políticas e avisos
          Nosso Portfólio Plus Icon Minus Icon
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline