Immunogen sequence: MEAIGHAGT CLGILANDGV LLAAERRNIH KLLDEVFFSE KIYKLNEDMA CSVAGITSDA NVLTNELRLI AQRYLLQYQE PIPCEQLVTA LCDIKQAYTQ FGGKRPFGVS LLYIGWDKHY GFQLYQSDPS GNYGGWKATC IGNNSAAAVS MLKQDYKEGE MTLKSALALA IKVLNKTMDV SKLSAEKVEI ATLTRENGKT VIRVLKQKEV EQLIKKHEEE EAKAEREKKE KEQKEKDK
The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the peptidase T1A family, that is a 20S core alpha subunit. Three alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Protein Aliases: alpha-3; Macropain subunit C9; MGC111191; MGC12467; MGC24813; Multicatalytic endopeptidase complex subunit C9; multicatalytic proteinase subunit L; proteasome (prosome, macropain) subunit, alpha type, 4; proteasome alpha 4 subunit; Proteasome component C9; proteasome subunit alpha 4; Proteasome subunit alpha type-4; Proteasome subunit alpha-3; proteasome subunit alpha3; proteasome subunit C9-like protein; proteasome subunit HC9; Proteasome subunit L; PSMA 4; unnamed protein product
Gene Aliases: C9; HC9; HsT17706; PSC9; PSMA4
UniProt ID: (Human) P25789, (Mouse) Q9R1P0, (Rat) P21670
Entrez Gene ID: (Rabbit) 100342159, (Pig) 397058, (Human) 5685, (Mouse) 26441, (Rat) 29671