Immunogen sequence: MSDIEEVVE EYEEEEQEEA AVEEQEEAAE EDAEAEAETE ETRAEEDEEE EEAKEAEDGP MEESKPKPRS FMPNLVPPKI PDGERVDFDD IHRKRMEKDL NELQALIEAH FENRKKEEEE LVSLKDRIER RRAERAEQQR IRNEREKERQ NRLAEERARR EEEENRRKAE DEARKKKALS NMMHFGGYIQ KTERKSGKRQ TEREKKKKIL AERRKVLAID HLNEDQLREK AKELWQSIYN LEAEKFDLQE KFKQQKYEIN VLRNRINDNQ KVSKTRGKAK VTGRWK
Cardiac Troponin T is the tropomyosin-binding subunit of the troponin complex, which is located on the thin filament of striated muscles and regulates muscle contraction in response to alterations in intracellular calcium ion concentration. Cardiac Troponin T is important in proper muscle function and myofibril formation. In the human heart, four cTnT isoforms are expressed in a developmentally regulated manner. Cardiac Troponin T expression in the human heart is affected by heart failure. Mutations within the Cardiac Troponin T protein have been associated with familial hypertrophic cardiomyopathy as well as with dilated cardiomyopathy. Transcripts for the Cardiac Troponin T gene undergo alternative splicing that results in many tissue-specific isoforms.
Protein Aliases: cardiac muscle troponin T; cardiac TnT; cardiac troponin T form I; cardiac troponin T isoform I; cardiac troponin T2; cTnT; MGC3889; tnTC; troponin T; Troponin T cardiac; troponin T type 2 (cardiac); Troponin T, cardiac muscle; Troponin T2; unnamed protein product
Gene Aliases: cTnT; Ctt; CTTG; RATCTTG; Tnnt3; Tnt
UniProt ID: (Chicken) P02642, (Rat) P50753, (Mouse) P50752
Entrez Gene ID: (Rabbit) 100009428, (Pig) 100622450, (Chicken) 396433, (Rat) 24837, (Mouse) 21956