    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • RNF31 Antibodies
          Proteintech
          RNF31/HOIP Recombinant Rabbit Monoclonal Antibody (230446H1), CoraLite® Plus 488
          View all (8) RNF31 antibodies
          Tech Support
          Cite RNF31/HOIP Recombinant Rabbit Monoclonal Antibody (230446H1), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          RNF31/HOIP Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          RNF31/HOIP Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          RNF31/HOIP Antibody (CL488-83137-3) in ICC/IF
          Immunofluorescent analysis of (-20°C Ethanol) fixed HepG2 cells using CoraLite® Plus 488 RNF31 / HOIP antibody (CL488-83137-3, Clone: 230446H1) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          RNF31/HOIP Antibody in Immunocytochemistry (ICC/IF)
          RNF31/HOIP Antibody in Flow Cytometry (Flow)
          RNF31/HOIP Recombinant Rabbit Monoclonal Antibody (230446H1), CoraLite® Plus 488
          Product Details
          CL488-83137-3
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Flow Cytometry (Flow)
          0.80 µg/10^6 cells in a 100 µL suspension
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230446H1
          Immunogen
          RNF31 / HOIP fusion protein Ag34505
          3D Epitope / Immunogen
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: RDCLFYLRDW TALRLQKLLQ DNNVMFNTEP PAGARAVPGG GCRVIEQKEV PNGLRDEACG KETPAGYAGL CQAHYKEYLV SLINAHSLDP ATLYEVEELE TATERYLHVR PQPLAGEDPP AYQARLLQKL TEEVPLGQSI PRRRK
          Target Information
          Lyn proto oncogene (Lcl/Yes related novel protein tyrosine kinase) is encoded by the Lyn gene located on chromosome 8 in humans and belongs to the Src family kinase. It is a non-receptor tyrosine kinase that acts as a mediator in transducing cellular signaling. The most well studied expression of Lyn is in hematopoietic cell types, both lymphoid and myeloid cells with exclusion in T lymphocytes. It has also been observed that Lyn plays diverse roles such as in regulation of B cell receptor signaling, mast cell signaling and Epithelial - Mesenchymal Transition (EMT).
          Bioinformatics
          Protein Aliases: E3 ubiquitin-protein ligase RNF31; FLJ23501; HOIL-1-interacting protein; HOIP; MGC19975; RING finger protein 31; RING-type E3 ubiquitin transferase RNF31; unnamed protein product; Zinc in-between-RING-finger ubiquitin-associated domain protein
          View more View less
          Gene Aliases: HOIP; IMD115; Paul; RNF31; ZIBRA
          View more View less
          UniProt ID: (Human) Q96EP0
          View more View less
          Entrez Gene ID: (Human) 55072
          View more View less
          Function(s)
          ubiquitin-protein transferase activity protein binding zinc ion binding transferase activity ubiquitin protein ligase binding K48-linked polyubiquitin binding identical protein binding ubiquitin binding metal ion binding ubiquitin protein ligase activity K63-linked polyubiquitin binding linear polyubiquitin binding
          Process(es)
          protein polyubiquitination cell surface receptor signaling pathway I-kappaB kinase/NF-kappaB signaling protein ubiquitination CD40 signaling pathway defense response to bacterium positive regulation of I-kappaB kinase/NF-kappaB signaling negative regulation of I-kappaB kinase/NF-kappaB signaling T cell receptor signaling pathway positive regulation of NF-kappaB transcription factor activity negative regulation of necroptotic process protein linear polyubiquitination positive regulation of protein targeting to mitochondrion positive regulation of xenophagy
          • News News
          • Responsibility Responsibility
          Chile flag icon
          Chile
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline