    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
          • Primary Antibodies ›
          • CD56 (NCAM) Antibodies
          Proteintech
          NCAM1/CD56 Recombinant Rabbit Monoclonal Antibody (240078C9), CoraLite® Plus 488
          View all (186) CD56 (NCAM) antibodies
          Tech Support
          Cite NCAM1/CD56 Recombinant Rabbit Monoclonal Antibody (240078C9), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          NCAM1/CD56 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          NCAM1/CD56 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          NCAM1/CD56 Antibody (CL488-83365-6) in ICC/IF
          Immunofluorescent analysis of (-20°C Ethanol) fixed SH-SY5Y cells using CoraLite Plus 488 NCAM1/CD56 antibody (CL488-83365-6, Clone: 240078C9 ) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          NCAM1/CD56 Antibody in Immunocytochemistry (ICC/IF)
          NCAM1/CD56 Recombinant Rabbit Monoclonal Antibody (240078C9), CoraLite® Plus 488
          Product Details
          CL488-83365-6
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:200
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          240078C9
          Immunogen
          Recombinant protein
          View immunogen
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • CoraLite 532
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: LQVDIVPSQG EISVGESKFF LCQVAGDAKD KDISWFSPNG EKLTPNQQRI SVVWNDDSSS TLTIYNANID DAGIYKCVVT GEDGSESEAT VNVKIFQKLM FKNAPTPQEF REGEDAVIVC DVVSSLPPTI IWKHKGRDVI LKKDVRFIVL SNNYLQIRGI KKTDEGTYRC EGRILARGEI NFKDIQVIVN VPPTIQARQN IVNATANLGQ SVTLVCDAEG FPEPTMSWTK DGEQIEQEED DEKYIFSDDS SQLTIKKVDK NDEAEYICIA ENKAGEQDAT IHLKVFAKPK ITYVENQTAM ELEEQVTLTC EASGDPIPSI TWRTSTRNIS SEEKASWTRP EKQETLDGHM VVRSHARVSS LTLKSIQYTD AGEYICTASN TIGQDSQSMY LEVQYAPKLQ GPVAVYTWEG NQVNITCEVF AYPSATISWF RDGQLLPSSN YSNIKIYNTP SASYLEVTPD SENDFGNYNC TAVNRIGQES LEFILVQADT PSSPSIDQVE PYSSTAQVQF DEPEATGGVP ILKYKAEWRA VGEEVWHSKW YDAKEASMEG IVTIVGLKPE TTYAVRLAAL NGKGLGEISA ASEFKTQPVQ GEPSAPKLEG QMGEDGNSIK VNLIKQDDGG SPIRHYLVRY RALSSEWKPE IRLPSGSDHV MLKSLDWNAE YEVYVVAENQ QGKSKAAHFV FRTSAQPTAI PANGSPTSG
          Target Information
          CD56, also known as neural cell adhesion molecule (NCAM), is a highly glycosylated transmembrane glycoprotein of the immunoglobulin family. It plays a crucial role in cell adhesion, migration, axonal growth, pathfinding, and synaptic plasticity. CD56 is ubiquitously expressed in the nervous system in isoforms ranging from 120-180 kDa and is involved in homotypic adhesion of neural cells. It mediates interactions by binding extracellular matrix components such as laminin and integrins, with polysialic modification reducing CD56-mediated adhesion. In the hematopoietic system, CD56 is expressed on natural killer (NK) cells and a subset of T cells known as NKT cells. It is also found on most neuroectodermal-derived cell lines, tissues, and neoplasms, including retinoblastoma, medulloblastoma, astrocytomas, and neuroblastoma. CD56 serves as a widely used neuroendocrine marker with high sensitivity for neuroendocrine tumors and ovarian granulosa cell tumors. Diseases associated with CD56 dysfunction include rabies and blastic plasmacytoid dendritic cell neoplasms, highlighting its importance in both neural and immune system functions.
          Bioinformatics
          Protein Aliases: 145 kda neural cell adhesion molecule; antigen recognized by monoclonal antibody 5.1H11; CD-56; CD56; E NCAM; membrane glycoprotein; N CAM1; N-CAM; N-CAM-1; neural cell adhesion molecule; Neural cell adhesion molecule 1; neural cell adhesion molecule secreted isoform; neural cell adhesion molecule, NCAM; sCD56; sNCAM; soluble CD56; soluble NCAM; unnamed protein product
          View more View less
          Gene Aliases: CD56; MSK39; NCAM; NCAM1
          View more View less
          UniProt ID: (Human) P13591
          View more View less
          Entrez Gene ID: (Human) 4684
          View more View less
          Function(s)
          virus receptor activity protein binding
          Process(es)
          epithelial to mesenchymal transition cell adhesion viral entry into host cell commissural neuron axon guidance regulation of semaphorin-plexin signaling pathway
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          India flag icon
          India
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline