    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
  • Services
    • 360° CDMO and CRO Solutions
          • Primary Antibodies ›
          • CD56 (NCAM) Antibodies
          Proteintech
          NCAM1/CD56 Recombinant Rabbit Monoclonal Antibody (240078C9), CoraLite® 532
          View all (145) CD56 (NCAM) antibodies
          Tech Support
          Cite NCAM1/CD56 Recombinant Rabbit Monoclonal Antibody (240078C9), CoraLite® 532
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          NCAM1/CD56 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          NCAM1/CD56 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          NCAM1/CD56 Antibody (CL532-83365-6) in ICC/IF
          Immunofluorescent analysis of (-20°C Ethanol) fixed SH-SY5Y cells using CoraLite532 NCAM1/CD56 antibody (CL532-83365-6, Clone: 240078C9 ) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          NCAM1/CD56 Antibody in Immunocytochemistry (ICC/IF)
          NCAM1/CD56 Recombinant Rabbit Monoclonal Antibody (240078C9), CoraLite® 532
          Product Details
          CL532-83365-6
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          240078C9
          Immunogen
          NCAM1/CD56 fusion protein Eg31816
          3D Epitope / Immunogen
          Conjugate
          CoraLite® 532 CoraLite® 532 CoraLite® 532
          • Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Excitation/Emission Max
          527/558 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: LQVDIVPSQG EISVGESKFF LCQVAGDAKD KDISWFSPNG EKLTPNQQRI SVVWNDDSSS TLTIYNANID DAGIYKCVVT GEDGSESEAT VNVKIFQKLM FKNAPTPQEF REGEDAVIVC DVVSSLPPTI IWKHKGRDVI LKKDVRFIVL SNNYLQIRGI KKTDEGTYRC EGRILARGEI NFKDIQVIVN VPPTIQARQN IVNATANLGQ SVTLVCDAEG FPEPTMSWTK DGEQIEQEED DEKYIFSDDS SQLTIKKVDK NDEAEYICIA ENKAGEQDAT IHLKVFAKPK ITYVENQTAM ELEEQVTLTC EASGDPIPSI TWRTSTRNIS SEEKASWTRP EKQETLDGHM VVRSHARVSS LTLKSIQYTD AGEYICTASN TIGQDSQSMY LEVQYAPKLQ GPVAVYTWEG NQVNITCEVF AYPSATISWF RDGQLLPSSN YSNIKIYNTP SASYLEVTPD SENDFGNYNC TAVNRIGQES LEFILVQADT PSSPSIDQVE PYSSTAQVQF DEPEATGGVP ILKYKAEWRA VGEEVWHSKW YDAKEASMEG IVTIVGLKPE TTYAVRLAAL NGKGLGEISA ASEFKTQPVQ GEPSAPKLEG QMGEDGNSIK VNLIKQDDGG SPIRHYLVRY RALSSEWKPE IRLPSGSDHV MLKSLDWNAE YEVYVVAENQ QGKSKAAHFV FRTSAQPTAI PANGSPTSG
          Target Information
          CD56, also known as neural cell adhesion molecule (NCAM), is a highly glycosylated transmembrane glycoprotein of the immunoglobulin family. It plays a crucial role in cell adhesion, migration, axonal growth, pathfinding, and synaptic plasticity. CD56 is ubiquitously expressed in the nervous system in isoforms ranging from 120-180 kDa and is involved in homotypic adhesion of neural cells. It mediates interactions by binding extracellular matrix components such as laminin and integrins, with polysialic modification reducing CD56-mediated adhesion. In the hematopoietic system, CD56 is expressed on natural killer (NK) cells and a subset of T cells known as NKT cells. It is also found on most neuroectodermal-derived cell lines, tissues, and neoplasms, including retinoblastoma, medulloblastoma, astrocytomas, and neuroblastoma. CD56 serves as a widely used neuroendocrine marker with high sensitivity for neuroendocrine tumors and ovarian granulosa cell tumors. Diseases associated with CD56 dysfunction include rabies and blastic plasmacytoid dendritic cell neoplasms, highlighting its importance in both neural and immune system functions.
          Bioinformatics
          Protein Aliases: 145 kda neural cell adhesion molecule; antigen recognized by monoclonal antibody 5.1H11; CD-56; CD56; E NCAM; membrane glycoprotein; N CAM1; N-CAM; N-CAM-1; neural cell adhesion molecule; Neural cell adhesion molecule 1; neural cell adhesion molecule secreted isoform; neural cell adhesion molecule, NCAM; sCD56; sNCAM; soluble CD56; soluble NCAM; unnamed protein product
          View more View less
          Gene Aliases: CD56; MSK39; NCAM; NCAM1
          View more View less
          UniProt ID: (Human) P13591
          View more View less
          Entrez Gene ID: (Human) 4684
          View more View less
          Function(s)
          virus receptor activity protein binding
          Process(es)
          epithelial to mesenchymal transition cell adhesion viral entry into host cell commissural neuron axon guidance regulation of semaphorin-plexin signaling pathway
          Hong Kong flag icon
          Hong Kong
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline