    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Calpain 2 Antibodies
          Proteintech
          CAPN2 Monoclonal Antibody (1E1F10), CoraLite® Plus 594
          View all (25) Calpain 2 antibodies
          Tech Support
          Cite CAPN2 Monoclonal Antibody (1E1F10), CoraLite® Plus 594
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          CAPN2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          CAPN2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          CAPN2 Antibody (CL594-66977) in ICC/IF
          Immunofluorescent analysis of (-20°C Ethanol) fixed HepG2 cells using CoraLite® Plus 594 CAPN2 antibody (CL594-66977, Clone: 1E1F10) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CAPN2 Antibody in Immunocytochemistry (ICC/IF)
          CAPN2 Monoclonal Antibody (1E1F10), CoraLite® Plus 594
          Product Details
          CL594-66977
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          1E1F10
          Immunogen
          CAPN2 fusion protein Ag28427
          3D Epitope / Immunogen
          Conjugate
          CoraLite® Plus 594 CoraLite® Plus 594 CoraLite® Plus 594
          • Unconjugated
          • CoraLite Plus 488
          • CoraLite Plus 647
          • Request custom conjugation
          Excitation/Emission Max
          588/604 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein G
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MAGIAAKLAK DREAAEGLGS HERAIKYLNQ DYEALRNECL EAGTLFQDPS FPAIPSALGF KELGPYSSKT RGIEWKRPTE ICADPQFIIG GATRTDICQG ALGDCWLLAA IASLTLNEEI LARVVPLNQS FQENYAGIFH FQFWQYGEWV EVVVDDRLPT KDGELLFVHS AEGSEFWSAL LEKAYAKING CYEALSGGAT TEGFEDFTGG IAEWYELKKP PPNLFKIIQK ALQKGSLLGC SIDITSAADS EAITFQKLVK GHAYSVTGAE EVESNGSLQK LIRIRNPWGE VEWTGRWNDN CPSWNTIDPE ERERLTRRHE DGEFWMSFSD
          Target Information
          Mu-Calpain, also known as Calpain-I, and m-calpain, also known as Calpain-II, are intracellular, calcium-dependent cysteine proteases. Mu-calpain has a micromolar sensitivity (thus the mu-) as compared to the millimolar calcium sensitivity of m-calpain.
          Bioinformatics
          Protein Aliases: 80 kDa M-calpain subunit; Ca2-activated neutral protease subunit; Calcium-activated neutral proteinase 2; CALP80; calpain 2, (m/II) large subunit; calpain 2, large [catalytic] subunit; calpain 2, large subunit; Calpain large polypeptide L2; Calpain M-type; calpain, large polypeptide L2; Calpain-2 catalytic subunit; Calpain-2 large subunit; CANP 2; cysteine proteinase; M-calpain; m-calpain 80 kDa large subunit; m-calpain large subunit; Millimolar-calpain; unnamed protein product
          View more View less
          Gene Aliases: AI326419; CANP2; CANPL2; CANPml; Capa-2; Capa2; CAPN2; m-calpain; m-calpin; mCANP
          View more View less
          UniProt ID: (Human) P17655, (Mouse) O08529, (Rat) Q07009
          View more View less
          Entrez Gene ID: (Human) 824, (Mouse) 12334, (Rat) 29154
          View more View less
          Function(s)
          calcium-dependent cysteine-type endopeptidase activity calcium ion binding protein binding cytoskeletal protein binding peptidase activity cysteine-type peptidase activity hydrolase activity enzyme binding macromolecular complex binding metal ion binding protein heterodimerization activity cysteine protease
          Process(es)
          response to hypoxia blastocyst development proteolysis myoblast fusion female pregnancy response to mechanical stimulus positive regulation of cardiac muscle cell apoptotic process protein autoprocessing protein catabolic process regulation of interleukin-6 production cellular response to interferon-beta response to hydrogen peroxide behavioral response to pain synaptic vesicle endocytosis regulation of cytoskeleton organization proteolysis involved in cellular protein catabolic process cellular response to lipopolysaccharide cellular response to amino acid stimulus vascular endothelial cell response to laminar fluid shear stress vascular endothelial cell response to oscillatory fluid shear stress protein catabolic process at postsynapse positive regulation of myoblast fusion positive regulation of phosphatidylcholine biosynthetic process
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline