    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
  • Services
    • 360° CDMO and CRO Solutions
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
          • Primary Antibodies ›
          • Human IgM Antibodies
          Proteintech
          IgM Monoclonal Antibody (2D10B10), CoraLite® Plus 647
          View all (70) Human IgM antibodies
          Tech Support
          Cite IgM Monoclonal Antibody (2D10B10), CoraLite® Plus 647
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          IgM Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          IgM Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          IgM Antibody (CL647-66484) in IHC (P)
          Immunofluorescent analysis of (4% PFA) fixed human appendicitis tissue using CoraLite Plus 647 human IgM antibody (CL647-66484, Clone: 2D10B10 ) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          IgM Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Product Details
          CL647-66484
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Paraffin) (IHC (P))
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          2D10B10
          Immunogen
          human IgM fusion protein Ag1459
          Conjugate
          CoraLite® Plus 647 CoraLite® Plus 647 CoraLite® Plus 647
          • Unconjugated
          • CoraLite 594
          • CoraLite Plus 488
          • HRP
          • Request custom conjugation
          Excitation/Emission Max
          654/674 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: FGNPRKSKLI CQATGFSPRQ IQVSWLREGK QVGSGVTTDQ VQAEAKESGP TTYKVTSTLT IKESDWLSQS MFTCRVDHRG LTFQQNASSM CVPDQDTAIR VFAIPPSFAS IFLTKSTKLT CLVTDLTTYD SVTISWTRQN GEAVKTHTNI SESHPNATFS AVGEASICED DWNSGERFTC TVTHTDLPSP LKQTISRPKG VALHRPDVYL LPPAREQLNL RESATITCLV TGFSPADVFV QWMQRGQPLS PEKYVTSAPM PEPQAPGRYF AHSILTVSEE EWNTGETYTC VVAHEALPNR VTERTVDKST EGEVSADEEG FENLWATAST FIVLFLLSLF YSTTVTLFKV K
          Target Information
          IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells.
          Bioinformatics
          Protein Aliases: belongs to immunoglobulin superfamily; Ig mu heavy chain constant region CH3 domain; unnamed protein product
          View more View less
          Gene Aliases: AGM1; MU; VH
          View more View less
          Entrez Gene ID: (Human) 3507
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline