    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
  • Services
    • 360° CDMO and CRO Solutions
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
          • Primary Antibodies ›
          • Human IgM Antibodies
          Proteintech
          IgM Monoclonal Antibody (2D10B10), HRP
          View all (71) Human IgM antibodies
          Tech Support
          Cite IgM Monoclonal Antibody (2D10B10), HRP
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          IgM Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          IgM Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          IgM Antibody (HRP-66484) in WB
          Various lysates were subjected to SDS PAGE followed by western blot with HRP-80713 (Beta Tubulin antibody) at dilution of 1:8,000 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          IgM Antibody in Western Blot (WB)
          Product Details
          HRP-66484
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:5,000-1:50,000
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          2D10B10
          Immunogen
          human IgM fusion protein Ag1459
          Conjugate
          HRP HRP HRP
          • Unconjugated
          • CoraLite 594
          • CoraLite Plus 488
          • CoraLite Plus 647
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1000 µg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: FGNPRKSKLI CQATGFSPRQ IQVSWLREGK QVGSGVTTDQ VQAEAKESGP TTYKVTSTLT IKESDWLSQS MFTCRVDHRG LTFQQNASSM CVPDQDTAIR VFAIPPSFAS IFLTKSTKLT CLVTDLTTYD SVTISWTRQN GEAVKTHTNI SESHPNATFS AVGEASICED DWNSGERFTC TVTHTDLPSP LKQTISRPKG VALHRPDVYL LPPAREQLNL RESATITCLV TGFSPADVFV QWMQRGQPLS PEKYVTSAPM PEPQAPGRYF AHSILTVSEE EWNTGETYTC VVAHEALPNR VTERTVDKST EGEVSADEEG FENLWATAST FIVLFLLSLF YSTTVTLFKV K
          Target Information
          IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells.
          Bioinformatics
          Protein Aliases: belongs to immunoglobulin superfamily; Ig mu heavy chain constant region CH3 domain; unnamed protein product
          View more View less
          Gene Aliases: AGM1; MU; VH
          View more View less
          Entrez Gene ID: (Human) 3507
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline