    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • XLF Antibodies
          Proteintech
          XLF Monoclonal Antibody (1F8C7), CoraLite® Plus 750
          View all (41) XLF antibodies
          Tech Support
          Cite XLF Monoclonal Antibody (1F8C7), CoraLite® Plus 750
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          XLF Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          XLF Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          XLF Antibody (CL750-66552) in WB
          Various lysates were subjected to SDS PAGE followed by western blot with CL750-66552 (XLF antibody) at dilution of 1:2,000 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          XLF Antibody in Western Blot (WB)
          Product Details
          CL750-66552
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000-1:4,000
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          1F8C7
          Immunogen
          XLF fusion protein Ag2496
          Conjugate
          CoraLite® Plus 750 CoraLite® Plus 750 CoraLite® Plus 750
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          755/780 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MEELEQGLL MQPWAWLQLA ENSLLAKVFI TKQGYALLVS DLQQVWHEQV DTSVVSQRAK ELNKRLTAPP AAFLCHLDNL LRPLLKDAAH PSEATFSCDC VADALILRVR SELSGLPFYW NFHCMLASPS LVSQHLIRPL MGMSLALQCQ VRELATLLHM KDLEIQDYQE SGATLIRDRL KTEPFEENSF LEQFMIEKLP EACSIGDGKP FVMNLQDLYM AVTTQEVQVG QKHQGAGDPH TSNSASLQGI DSQCVNQPEQ LVSSAPTLSA PEKESTGTSG PLQRPQLSKV KRKKPRGLFS
          Target Information
          Double-strand breaks in DNA result from genotoxic stresses and are among the most damaging of DNA lesions. This gene encodes a DNA repair factor essential for the nonhomologous end-joining pathway, which preferentially mediates repair of double-stranded breaks. Mutations in this gene cause different kinds of severe combined immunodeficiency disorders.
          Bioinformatics
          Protein Aliases: FLJ12610; Non-homologous end-joining factor 1; nonhomologous end-joining factor 1; Protein cernunnos; unnamed protein product; XRCC4-like factor
          View more View less
          Gene Aliases: IMD124; MCOPCB13; NHEJ1; XLF
          View more View less
          UniProt ID: (Human) Q9H9Q4
          View more View less
          Entrez Gene ID: (Human) 79840
          View more View less
          Function(s)
          DNA binding protein binding DNA end binding DNA polymerase binding
          Process(es)
          telomere maintenance DNA repair double-strand break repair double-strand break repair via nonhomologous end joining cellular response to DNA damage stimulus central nervous system development response to ionizing radiation B cell differentiation T cell differentiation immunoglobulin V(D)J recombination positive regulation of ligase activity
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline