    • See all product categories
  • Applications
    • Cell Analysis
    • Protein Biology
    • Lab Informatics
    • 360° CDMO and CRO Solutions
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
          • Primary Antibodies ›
          • RILP Antibodies
          Proteintech
          RILP Recombinant Rabbit Monoclonal Antibody (230208G2), CoraLite® Plus 750
          View all (3) RILP antibodies
          Tech Support
          Cite RILP Recombinant Rabbit Monoclonal Antibody (230208G2), CoraLite® Plus 750
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          RILP Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          RILP Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          RILP Antibody (CL750-82996-4) in WB
          0.1 μL human plasma was subjected to SDS PAGE followed by western blot with HRP-66484 (HRP conjugated mouse anti-human IgM) at dilution of 1:10,000, 1:20,000, 1:40,000, incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          RILP Antibody in Western Blot (WB)
          RILP Recombinant Rabbit Monoclonal Antibody (230208G2), CoraLite® Plus 750
          Product Details
          CL750-82996-4
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:1,000
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230208G2
          Immunogen
          RILP fusion protein Ag4472
          3D Epitope / Immunogen
          Conjugate
          CoraLite® Plus 750 CoraLite® Plus 750 CoraLite® Plus 750
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          755/780 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: ELRAGPQEER ALLRQLKEVT DRQRDELRAH NRDLRQRGQE TEALQEQLQR LLLVNAELRH KLAAMQTQLR AAQDRERERQ QPGEAATPQA KERARGQAGR PGHQHGQEPE WATAGAGAPG NPEDPAEAAQ QLGRPSEAGQ CRFSREEFEQ ILQERNELKA KVFLLKEELA YFQRELLTDH RVPGLLLEAM KVAVRKQRKK IKAKMLGTPE EAESSEDEAG PWILLSDDKG DHPPPPESKI QSFFGLWYRG KAESSEDETS SPAPSKLGGE EEAQPQSPAP DPPCSALHEH LCLGASAAPE A
          Target Information
          The Rab interacting lysosomal protein (RILP) is lysosomal protein that interacts with RAB7, a small GTPase that controls transport to endocytic degradative compartments. It is thought that RILP is a downstream effector of RAB7 and both proteins act together in the regulation of late endocytic traffic. Overexpression of RILP caused enlarged lysosomes that are more centrally located in the cell suggesting that RILP may also be involved in the regulation of lysosomal morphology. Other evidence suggests that RILP may play a role in the biogenesis of multivesicular bodies.
          Bioinformatics
          Protein Aliases: Rab-interacting lysosomal protein; unnamed protein product
          View more View less
          Gene Aliases: PP10141; RILP
          View more View less
          UniProt ID: (Human) Q96NA2
          View more View less
          Entrez Gene ID: (Human) 83547
          View more View less
          Function(s)
          protein binding small GTPase binding protein dimerization activity dynein light intermediate chain binding
          Process(es)
          endosome to lysosome transport protein transport endosome transport via multivesicular body sorting pathway negative regulation of protein catabolic process early endosome to late endosome transport positive regulation of protein catabolic process cilium morphogenesis intralumenal vesicle formation
          • Trademarks
          • Fair Trade Fair Trade
          Korea flag icon
          Korea
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910
          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783
          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)
          ISMS Logo
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline