          • Primary Antibodies ›
          • RILP Antibodies
          Proteintech
          RILP Recombinant Rabbit Monoclonal Antibody (230208G2)
          View all (3) RILP antibodies
          Tech Support
          Cite RILP Recombinant Rabbit Monoclonal Antibody (230208G2)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (4)
          RILP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          RILP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          RILP Antibody (82996-4-RR) in IHC (P)
          Immunohistochemical analysis of paraffin-embedded human ovarian cancer slide using 82996-4-RR (RILP antibody) at dilution of 1:500 (under 20x lens). Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          RILP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RILP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RILP Antibody in Western Blot (WB)
          RILP Antibody in Surface plasmon resonance (SPR)
          RILP Recombinant Rabbit Monoclonal Antibody (230208G2)
          Product Details
          82996-4-RR
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000-1:4,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:500
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Surface plasmon resonance (SPR)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          CHO cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230208G2
          Immunogen
          RILP fusion protein Ag4472
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 750
          • Request custom conjugation
          Form
          Liquid
          Concentration
          0.8 mg/mL
          Amount
          80 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: ELRAGPQEER ALLRQLKEVT DRQRDELRAH NRDLRQRGQE TEALQEQLQR LLLVNAELRH KLAAMQTQLR AAQDRERERQ QPGEAATPQA KERARGQAGR PGHQHGQEPE WATAGAGAPG NPEDPAEAAQ QLGRPSEAGQ CRFSREEFEQ ILQERNELKA KVFLLKEELA YFQRELLTDH RVPGLLLEAM KVAVRKQRKK IKAKMLGTPE EAESSEDEAG PWILLSDDKG DHPPPPESKI QSFFGLWYRG KAESSEDETS SPAPSKLGGE EEAQPQSPAP DPPCSALHEH LCLGASAAPE A
          Target Information
          The Rab interacting lysosomal protein (RILP) is lysosomal protein that interacts with RAB7, a small GTPase that controls transport to endocytic degradative compartments. It is thought that RILP is a downstream effector of RAB7 and both proteins act together in the regulation of late endocytic traffic. Overexpression of RILP caused enlarged lysosomes that are more centrally located in the cell suggesting that RILP may also be involved in the regulation of lysosomal morphology. Other evidence suggests that RILP may play a role in the biogenesis of multivesicular bodies.
          Bioinformatics
          Protein Aliases: Rab-interacting lysosomal protein; unnamed protein product
          View more View less
          Gene Aliases: PP10141; RILP
          View more View less
          UniProt ID: (Human) Q96NA2
          View more View less
          Entrez Gene ID: (Human) 83547
          View more View less
          Function(s)
          protein binding small GTPase binding protein dimerization activity dynein light intermediate chain binding
          Process(es)
          endosome to lysosome transport protein transport endosome transport via multivesicular body sorting pathway negative regulation of protein catabolic process early endosome to late endosome transport positive regulation of protein catabolic process cilium morphogenesis intralumenal vesicle formation
          Contact tech support
          オンラインオーダー Plus Icon Minus Icon
          • オンラインオーダー可能な製品
          • オンラインオーダーに関する個人情報の取扱いについて
          • 特定商取引法に基づく表記
          • 返品について
          • 表示価格について
          • 配送料について
          サポート Plus Icon Minus Icon
          • FAQ、サポート情報
          • お問い合わせ一覧
          • サポートセンター
          • ドキュメント、証明書
          • 研究情報公開
          • Webサイトの問題を報告​
          リソース Plus Icon Minus Icon
          • ラーニングセンター
          • キャンペーン
          • イベント、セミナー
          • Blog
          • 企業情報
          • Social Responsibility (CSR)
          • お客さま体験の向上
          • 採用情報 採用情報
          • ニュース
          • 商標
          • ポリシーと通知
          • PPD
          • 標準販売条件
          • 個人情報保護方針
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline