Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: RGLAPQNKPE LQKVMEKRKR DQVIKQKEEE AQKKKSDLEI ELLKRQQKLE QLELEKQKLQ EEQENAPEFV KVKGNLRRTG QEVAQAQES
FAM107B is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C10orf45; FAM107B/CDNF fusion; FLJ45505; heat shock-inducible tumor small protein; HITS; MGC11034; novel DUF1151 domain containing protein; Protein FAM107B; unnamed protein product
Gene Aliases: 3110001A13Rik; AA589595; AA959793; AI132312; C10orf45; FAM107B; HITS
UniProt ID: (Human) Q9H098, (Mouse) Q3TGF2
Entrez Gene ID: (Human) 83641, (Mouse) 66540
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support