Search
Search
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGTQDPGNMG TGVPASEQIS CAKEDPQVYC PEETGGTKDV QVTDCKSPED SRPPKETDCC NPEDSGQLMV SYEGKAMGYQ VPPFGWRICL AHEFTEKRKP FQANNVSLSN MIKHIGMGLR YLQWWYRKTH VEKKTPFIDM INSVPLRQIY GCPLGGIGGG TITRGWRGQF CRWQLNPGMY QHRTVIADQF TVCLRREGQT VYQQVLSLER PSVLRSWNWG LCGYFAFYHA LYPRAWTVYQ LPGQNVTLTC RQITPILPHD YQDSSLPVGV FVWDVENEGD EALDVSIMFS MRNGLGGGDD APGGLWNEPF CLERSGETVR GLLLHHPTLP NPYTMAVAAR VTAATTVTHI TAFDPDSTGQ QVWQDLLQDG QLDSPTGQST PTQKGVGIAG AVCVSSKLRP RGQCRLEFSL AWDMPRIMFG AKGQVHYRRY TRFFGQDGDA APALSHYALC RYAEWEERIS AWQSPVLDDR SLPAWYKSAL FNELYFLADG GTVWLEVLED SLPEELGRNM CHLRPTLRDY GRFGYLEGQE YRMYNTYDVH FYASFALIML WPKLELSLQY DMALATLRED LTRRRYLMSG VMAPVKRRNV IPHDIGDPDD EPWLRVNAYL IHDTADWKDL NLKFVLQVYR DYYLTGDQNF LKDMWPVCLA VMESEMKFDK DHDGLIENGG YADQTYDGWV TTGPSAYCGG LWLAAVAVMV QMAALCGAQD IQDKFSSILS RGQEAYERLL WNGRYYNYDS SSRPQSRSVM SDQCAGQWFL KACGLGEGDT EVFPTQHVVR ALQTIFELNV QAFAGGAMGA VNGMQPHGVP DKSSVQSDEV WVGVVYGLAA TMIQEGLTWE GFQTAEGCYR TVWERLGLAF QTPEAYCQQR VFRSLAYMRP LSIWAMQLAL QQQQHKKASW PKVKQGTGLR TGPMFGPKEA MANLSPE
Non-lysosomal glucosylceramidase that catalyzes the conversion of glucosylceramide (GlcCer) to free glucose and ceramide. Involved in sphingomyelin generation and prevention of glycolipid accumulation. May also catalyze the hydrolysis of bile acid 3-O-glucosides, however, the relevance of such activity is unclear in vivo. Plays a role in central nevous system development. Required for proper formation of motor neuron axons.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3.2.1.45; beta-glucocerebrosidase 2; bile acid beta-glucosidase GBA2; bile acid glucosyl transferase GBA2; cholesterol glucosyltransferase GBA2; cholesteryl-beta-glucosidase GBA2; glucocerebrosidase 2; glucosidase, beta (bile acid) 2; glucosylceramidase 2; non-lysosomal cholesterol glycosyltransferase; non-lysosomal galactosylceramidase; non-lysosomal glycosylceramidase; unnamed protein product
Gene Aliases: AD035; NLGase; SPG46
UniProt ID: (Human) Q9HCG7
Entrez Gene ID: (Human) 57704
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support