Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MAERAALEE LVKLQGERVR GLKQQKASAE LIEEEVAKLL KLKAQLGPDE SKQKFVLKTP KGTRDYSPRQ MAVREKVFDV IIRCFKRHGA EVIDTPVFEL KETLMGKYGE DSKLIYDLKD QGGELLSLRY DLTVPFARYL AMNKLTNIKR YHIAKVYRRD NPAMTRGRYR EFYQCDFDIA GNFDPMIPDA ECLKIMCEIL SSLQIGDFLV KVNDRRILDG MFAICGVSDS KFRTICSSVD KLDKVSWEEV KNEMVGEKGL APEVADRIGD YVQQHGGVSL VEQ (1-282 aa encoded by BC011807)
Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. The protein encoded by this gene is a cytoplasmic enzyme which belongs to the class II family of aminoacyl-tRNA synthetases. The enzyme is responsible for the synthesis of histidyl-transfer RNA, which is essential for the incorporation of histidine into proteins. The gene is located in a head-to-head orientation with HARSL on chromosome five, where the homologous genes share a bidirectional promoter. The gene product is a frequent target of autoantibodies in the human autoimmune disease polymyositis/dermatomyositis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dead end homolog 1; FLJ20491; HisRS; histidine translase; histidine tRNA ligase; histidyl-tRNA synthetase, cytoplasmic; Jo-1; Jo-1 antigen; unnamed protein product
Gene Aliases: CMT2W; Dnd1; HARS; HRS; MMHRS; USH3B
UniProt ID: (Human) P12081, (Mouse) Q61035
Entrez Gene ID: (Human) 3035, (Rat) 307492, (Mouse) 15115
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support